Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28574585-28574828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDOC
Entrez Gene ID
230
Gene Name
aldolase, fructose-bisphosphate C
Gene Aliases
ALDC
UniGene
Hs.155247
Gene Chromosome Location
Chr.17: 28573115-28576963 on Build GRCh38
Transcripts
NM_005165.2 XM_005257949.2 XM_011524556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4579549 c.501C>T p.N167N Chr.17: 28574735-28574735 on Build GRCh38
4607657 c.534C>A p.C178* Chr.17: 28574702-28574702 on Build GRCh38
5526517 c.479C>T p.S160F Chr.17: 28574757-28574757 on Build GRCh38
2800344 c.446G>T p.R149L Chr.17: 28574790-28574790 on Build GRCh38
271004 c.517C>T p.R173C Chr.17: 28574719-28574719 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754828649 Chr.17: 28574780-28574780 on Build GRCh38
rs747367976 Chr.17: 28574719-28574719 on Build GRCh38
rs755096574 Chr.17: 28574693-28574693 on Build GRCh38
rs758665617 Chr.17: 28574715-28574715 on Build GRCh38
rs199611960 Chr.17: 28574770-28574770 on Build GRCh38
rs751238851 Chr.17: 28574629-28574629 on Build GRCh38
rs368999434 Chr.17: 28574650-28574650 on Build GRCh38
rs915922016 Chr.17: 28574631-28574631 on Build GRCh38
rs753831994 Chr.17: 28574769-28574769 on Build GRCh38
rs778804657 Chr.17: 28574706-28574706 on Build GRCh38
rs752807581 Chr.17: 28574790-28574790 on Build GRCh38
rs200821079 Chr.17: 28574735-28574735 on Build GRCh38
rs920977425 Chr.17: 28574634-28574634 on Build GRCh38
rs767141084 Chr.17: 28574763-28574763 on Build GRCh38
rs369588229 Chr.17: 28574785-28574785 on Build GRCh38
rs148239543 Chr.17: 28574718-28574718 on Build GRCh38
rs748135082 Chr.17: 28574743-28574743 on Build GRCh38
rs771544765 Chr.17: 28574607-28574607 on Build GRCh38
rs185937578 Chr.17: 28574644-28574644 on Build GRCh38
rs763447848 Chr.17: 28574624-28574624 on Build GRCh38
rs201385668 Chr.17: 28574730-28574730 on Build GRCh38
rs756100602 Chr.17: 28574671-28574671 on Build GRCh38
rs753648020 Chr.17: 28574682-28574682 on Build GRCh38
rs771634478 Chr.17: 28574724-28574724 on Build GRCh38
rs750196586 Chr.17: 28574669-28574669 on Build GRCh38
rs373220558 Chr.17: 28574729-28574729 on Build GRCh38
rs748408664 Chr.17: 28574708-28574708 on Build GRCh38
rs762521807 Chr.17: 28574617-28574617 on Build GRCh38
rs767339232 Chr.17: 28574657-28574657 on Build GRCh38
rs774924303 Chr.17: 28574616-28574616 on Build GRCh38
rs766028700 Chr.17: 28574768-28574768 on Build GRCh38
rs558273823 Chr.17: 28574731-28574731 on Build GRCh38
rs758753501 Chr.17: 28574795-28574795 on Build GRCh38
rs745839133 Chr.17: 28574659-28574659 on Build GRCh38
rs948662159 Chr.17: 28574648-28574648 on Build GRCh38
rs142754133 Chr.17: 28574767-28574767 on Build GRCh38
rs780034115 Chr.17: 28574680-28574680 on Build GRCh38
rs773057473 Chr.17: 28574766-28574766 on Build GRCh38
rs899450951 Chr.17: 28574666-28574666 on Build GRCh38
rs190821159 Chr.17: 28574728-28574728 on Build GRCh38

View Full Product Details