Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 56192902-56193127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSCAN5B
Entrez Gene ID
342933
Gene Name
zinc finger and SCAN domain containing 5B
Gene Aliases
ZNF371, ZNF495B
UniGene
Hs.132130
Gene Chromosome Location
Chr.19: 56189689-56197890 on Build GRCh38
Transcripts
NM_001080456.2 XM_005258864.3 XM_006723189.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3539973 c.90A>T p.E30D Chr.19: 56192963-56192963 on Build GRCh38
2970442 c.24A>G p.S8S Chr.19: 56193029-56193029 on Build GRCh38
3539975 c.56G>A p.G19E Chr.19: 56192997-56192997 on Build GRCh38
2970438 c.71G>A p.R24Q Chr.19: 56192982-56192982 on Build GRCh38
4657790 c.148T>C p.F50L Chr.19: 56192905-56192905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760539327 Chr.19: 56193045-56193045 on Build GRCh38
rs752967962 Chr.19: 56192998-56192998 on Build GRCh38
rs776964901 Chr.19: 56192927-56192927 on Build GRCh38
rs766142573 Chr.19: 56192926-56192926 on Build GRCh38
rs764770107 Chr.19: 56193073-56193073 on Build GRCh38
rs759081899 Chr.19: 56193063-56193063 on Build GRCh38
rs139794340 Chr.19: 56192972-56192972 on Build GRCh38
rs575451380 Chr.19: 56193087-56193087 on Build GRCh38
rs765923750 Chr.19: 56192940-56192940 on Build GRCh38
rs368861394 Chr.19: 56192942-56192942 on Build GRCh38
rs368732521 Chr.19: 56192982-56192982 on Build GRCh38
rs767503640 Chr.19: 56192937-56192937 on Build GRCh38
rs80228467 Chr.19: 56192997-56192997 on Build GRCh38
rs778377838 Chr.19: 56192970-56192970 on Build GRCh38
rs950612646 Chr.19: 56193051-56193051 on Build GRCh38
rs753351356 Chr.19: 56192990-56192990 on Build GRCh38
rs555514356 Chr.19: 56193058-56193058 on Build GRCh38
rs767211827 Chr.19: 56193036-56193036 on Build GRCh38
rs758761780 Chr.19: 56193002-56193002 on Build GRCh38
rs551826201 Chr.19: 56193013-56193013 on Build GRCh38
rs200257976 Chr.19: 56192945-56192945 on Build GRCh38
rs535628209 Chr.19: 56193057-56193057 on Build GRCh38
rs374507565 Chr.19: 56192947-56192947 on Build GRCh38
rs773169405 Chr.19: 56192986-56192986 on Build GRCh38
rs776907331 Chr.19: 56192973-56192973 on Build GRCh38
rs528776961 Chr.19: 56192994-56192994 on Build GRCh38
rs763293023 Chr.19: 56193031-56193032 on Build GRCh38
rs539231465 Chr.19: 56193086-56193086 on Build GRCh38
rs771889363 Chr.19: 56193037-56193037 on Build GRCh38
rs1017313429 Chr.19: 56193008-56193008 on Build GRCh38
rs1007247006 Chr.19: 56193005-56193005 on Build GRCh38
rs757903505 Chr.19: 56193020-56193020 on Build GRCh38
rs761690422 Chr.19: 56192983-56192983 on Build GRCh38
rs760588021 Chr.19: 56192987-56192987 on Build GRCh38
rs752375513 Chr.19: 56193078-56193078 on Build GRCh38
rs571770602 Chr.19: 56193015-56193015 on Build GRCh38
rs772948603 Chr.19: 56193038-56193038 on Build GRCh38
rs763964002 Chr.19: 56192931-56192931 on Build GRCh38
rs781350323 Chr.19: 56193021-56193021 on Build GRCh38
rs919007729 Chr.19: 56193041-56193041 on Build GRCh38
rs61745809 Chr.19: 56193030-56193030 on Build GRCh38
rs745957412 Chr.19: 56193025-56193025 on Build GRCh38
rs759883974 Chr.19: 56192929-56192929 on Build GRCh38
rs749851397 Chr.19: 56192938-56192938 on Build GRCh38
rs770091536 Chr.19: 56193029-56193029 on Build GRCh38
rs377038668 Chr.19: 56192932-56192932 on Build GRCh38
rs755679725 Chr.19: 56192939-56192939 on Build GRCh38
rs372790304 Chr.19: 56193011-56193011 on Build GRCh38
rs761588691 Chr.19: 56192935-56192935 on Build GRCh38
rs779182763 Chr.19: 56192943-56192943 on Build GRCh38
rs887059907 Chr.19: 56193082-56193082 on Build GRCh38
rs758787037 Chr.19: 56192953-56192953 on Build GRCh38
rs201784103 Chr.19: 56192941-56192941 on Build GRCh38
rs765870687 Chr.19: 56192989-56192989 on Build GRCh38
rs931207459 Chr.19: 56193059-56193059 on Build GRCh38
rs770140640 Chr.19: 56192978-56192978 on Build GRCh38
rs1047435925 Chr.19: 56193062-56193062 on Build GRCh38
rs749510969 Chr.19: 56193036-56193036 on Build GRCh38

View Full Product Details