Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 20313493-20313722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNB1IP1
Entrez Gene ID
57820
Gene Name
cyclin B1 interacting protein 1
Gene Aliases
C14orf18, HEI10
UniGene
Hs.107003
Gene Chromosome Location
Chr.14: 20311368-20333312 on Build GRCh38
Transcripts
NM_021178.4 NM_182849.2 NM_182852.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5980538 c.559A>G p.T187A Chr.14: 20313540-20313540 on Build GRCh38
3744254 c.503A>G p.N168S Chr.14: 20313596-20313596 on Build GRCh38
6053977 c.379C>T p.Q127* Chr.14: 20313720-20313720 on Build GRCh38
3814414 c.416T>C p.M139T Chr.14: 20313683-20313683 on Build GRCh38
4955548 c.578G>T p.G193V Chr.14: 20313521-20313521 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752672389 Chr.14: 20313619-20313619 on Build GRCh38
rs142847094 Chr.14: 20313605-20313605 on Build GRCh38
rs767459771 Chr.14: 20313533-20313533 on Build GRCh38
rs762581759 Chr.14: 20313601-20313601 on Build GRCh38
rs796743555 Chr.14: 20313618-20313618 on Build GRCh38
rs747333513 Chr.14: 20313590-20313590 on Build GRCh38
rs772896629 Chr.14: 20313600-20313600 on Build GRCh38
rs372430614 Chr.14: 20313540-20313540 on Build GRCh38
rs764240856 Chr.14: 20313588-20313591 on Build GRCh38
rs139737493 Chr.14: 20313607-20313607 on Build GRCh38
rs1034750512 Chr.14: 20313596-20313596 on Build GRCh38
rs754084336 Chr.14: 20313644-20313644 on Build GRCh38
rs1021892128 Chr.14: 20313688-20313688 on Build GRCh38
rs371403814 Chr.14: 20313566-20313566 on Build GRCh38
rs745963898 Chr.14: 20313572-20313572 on Build GRCh38
rs868287069 Chr.14: 20313568-20313569 on Build GRCh38
rs760441303 Chr.14: 20313624-20313624 on Build GRCh38
rs762673575 Chr.14: 20313643-20313645 on Build GRCh38
rs369736561 Chr.14: 20313616-20313616 on Build GRCh38
rs193198359 Chr.14: 20313519-20313519 on Build GRCh38
rs750420914 Chr.14: 20313693-20313693 on Build GRCh38
rs776869250 Chr.14: 20313632-20313632 on Build GRCh38
rs765853177 Chr.14: 20313521-20313521 on Build GRCh38
rs375327618 Chr.14: 20313568-20313568 on Build GRCh38
rs1055983389 Chr.14: 20313610-20313610 on Build GRCh38
rs146791686 Chr.14: 20313593-20313593 on Build GRCh38
rs751513008 Chr.14: 20313527-20313527 on Build GRCh38
rs373325154 Chr.14: 20313663-20313663 on Build GRCh38
rs922235519 Chr.14: 20313537-20313537 on Build GRCh38
rs763936427 Chr.14: 20313629-20313629 on Build GRCh38
rs376980309 Chr.14: 20313655-20313655 on Build GRCh38
rs143558939 Chr.14: 20313548-20313548 on Build GRCh38
rs772648834 Chr.14: 20313580-20313580 on Build GRCh38
rs780728859 Chr.14: 20313585-20313585 on Build GRCh38
rs777941048 Chr.14: 20313543-20313543 on Build GRCh38
rs139806744 Chr.14: 20313528-20313528 on Build GRCh38

View Full Product Details