Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 90288035-90288308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRDE2
Entrez Gene ID
55051
Gene Name
NRDE-2, necessary for RNA interference, domain containing
Gene Aliases
C14orf102
UniGene
Hs.528131
Gene Chromosome Location
Chr.14: 90274276-90332137 on Build GRCh38
Transcripts
NM_017970.3 XM_011536886.1 XM_017021394.1 XM_017021395.1 XM_017021396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5350230 c.3158+9A>T p.? Chr.14: 90288208-90288208 on Build GRCh38
2250908 c.3125A>G p.K1042R Chr.14: 90288250-90288250 on Build GRCh38
6356849 c.3112A>G p.I1038V Chr.14: 90288263-90288263 on Build GRCh38
2250909 c.3106T>C p.F1036L Chr.14: 90288269-90288269 on Build GRCh38
5812998 c.3122A>T p.E1041V Chr.14: 90288253-90288253 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753642420 Chr.14: 90288186-90288186 on Build GRCh38
rs751681290 Chr.14: 90288238-90288238 on Build GRCh38
rs759260251 Chr.14: 90288219-90288219 on Build GRCh38
rs144099237 Chr.14: 90288092-90288092 on Build GRCh38
rs932715945 Chr.14: 90288065-90288065 on Build GRCh38
rs548572685 Chr.14: 90288138-90288138 on Build GRCh38
rs777517900 Chr.14: 90288286-90288286 on Build GRCh38
rs775693235 Chr.14: 90288223-90288223 on Build GRCh38
rs989423071 Chr.14: 90288140-90288140 on Build GRCh38
rs530990459 Chr.14: 90288185-90288185 on Build GRCh38
rs539682224 Chr.14: 90288212-90288212 on Build GRCh38
rs377721556 Chr.14: 90288069-90288069 on Build GRCh38
rs1030047493 Chr.14: 90288117-90288117 on Build GRCh38
rs776359689 Chr.14: 90288216-90288216 on Build GRCh38
rs763551783 Chr.14: 90288230-90288230 on Build GRCh38
rs999365255 Chr.14: 90288126-90288126 on Build GRCh38
rs1008681308 Chr.14: 90288091-90288091 on Build GRCh38
rs930089072 Chr.14: 90288195-90288195 on Build GRCh38
rs59075428 Chr.14: 90288070-90288070 on Build GRCh38
rs560254766 Chr.14: 90288203-90288203 on Build GRCh38
rs900199794 Chr.14: 90288105-90288105 on Build GRCh38
rs553679994 Chr.14: 90288176-90288176 on Build GRCh38
rs571006135 Chr.14: 90288204-90288204 on Build GRCh38
rs767729512 Chr.14: 90288249-90288249 on Build GRCh38
rs17799226 Chr.14: 90288194-90288194 on Build GRCh38
rs756308982 Chr.14: 90288279-90288279 on Build GRCh38
rs982527053 Chr.14: 90288182-90288182 on Build GRCh38
rs754699660 Chr.14: 90288188-90288188 on Build GRCh38
rs752395485 Chr.14: 90288284-90288284 on Build GRCh38
rs191120303 Chr.14: 90288285-90288285 on Build GRCh38
rs377518843 Chr.14: 90288167-90288167 on Build GRCh38
rs891094205 Chr.14: 90288079-90288079 on Build GRCh38
rs553168136 Chr.14: 90288233-90288233 on Build GRCh38
rs770338537 Chr.14: 90288220-90288220 on Build GRCh38
rs560473804 Chr.14: 90288111-90288111 on Build GRCh38
rs945603736 Chr.14: 90288277-90288277 on Build GRCh38
rs748167823 Chr.14: 90288189-90288189 on Build GRCh38
rs568362499 Chr.14: 90288156-90288156 on Build GRCh38
rs916521593 Chr.14: 90288154-90288154 on Build GRCh38
rs1037315796 Chr.14: 90288096-90288096 on Build GRCh38
rs1031287883 Chr.14: 90288127-90288127 on Build GRCh38
rs529218410 Chr.14: 90288123-90288123 on Build GRCh38
rs111746887 Chr.14: 90288077-90288077 on Build GRCh38
rs777373808 Chr.14: 90288200-90288200 on Build GRCh38
rs146015101 Chr.14: 90288263-90288263 on Build GRCh38
rs185775273 Chr.14: 90288110-90288110 on Build GRCh38
rs761822772 Chr.14: 90288246-90288246 on Build GRCh38

View Full Product Details