Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 78200106-78200357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP35
Entrez Gene ID
57558
Gene Name
ubiquitin specific peptidase 35
Gene Aliases
-
UniGene
Hs.531249
Gene Chromosome Location
Chr.11: 78188812-78233651 on Build GRCh38
Transcripts
NM_020798.2 XM_011545187.2 XM_011545188.2 XM_017018052.1 XM_017018053.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1357159 c.973G>A p.A325T Chr.11: 78200169-78200169 on Build GRCh38
932254 c.1024G>A p.E342K Chr.11: 78200220-78200220 on Build GRCh38
7078157 c.1032C>A p.F344L Chr.11: 78200228-78200228 on Build GRCh38
6818641 c.967C>T p.R323W Chr.11: 78200163-78200163 on Build GRCh38
1127631 c.1032C>T p.F344F Chr.11: 78200228-78200228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148081303 Chr.11: 78200164-78200164 on Build GRCh38
rs563421356 Chr.11: 78200163-78200163 on Build GRCh38
rs760223313 Chr.11: 78200241-78200242 on Build GRCh38
rs188252109 Chr.11: 78200301-78200301 on Build GRCh38
rs867404 Chr.11: 78200330-78200330 on Build GRCh38
rs777188359 Chr.11: 78200267-78200267 on Build GRCh38
rs758206967 Chr.11: 78200145-78200145 on Build GRCh38
rs763720231 Chr.11: 78200277-78200301 on Build GRCh38
rs547529805 Chr.11: 78200229-78200229 on Build GRCh38
rs562205827 Chr.11: 78200192-78200192 on Build GRCh38
rs1008822895 Chr.11: 78200314-78200314 on Build GRCh38
rs779173804 Chr.11: 78200138-78200138 on Build GRCh38
rs766961392 Chr.11: 78200193-78200193 on Build GRCh38
rs766513046 Chr.11: 78200128-78200128 on Build GRCh38
rs755648903 Chr.11: 78200231-78200231 on Build GRCh38
rs867405 Chr.11: 78200326-78200326 on Build GRCh38
rs769385250 Chr.11: 78200169-78200169 on Build GRCh38
rs764270200 Chr.11: 78200282-78200282 on Build GRCh38
rs748919914 Chr.11: 78200248-78200248 on Build GRCh38
rs781090025 Chr.11: 78200228-78200228 on Build GRCh38
rs765443661 Chr.11: 78200210-78200210 on Build GRCh38
rs772505617 Chr.11: 78200181-78200181 on Build GRCh38
rs866547145 Chr.11: 78200140-78200140 on Build GRCh38
rs771624107 Chr.11: 78200266-78200266 on Build GRCh38
rs777496605 Chr.11: 78200215-78200215 on Build GRCh38
rs371278103 Chr.11: 78200177-78200177 on Build GRCh38
rs757230311 Chr.11: 78200223-78200223 on Build GRCh38
rs527799146 Chr.11: 78200219-78200219 on Build GRCh38
rs2101309 Chr.11: 78200292-78200292 on Build GRCh38
rs529623459 Chr.11: 78200126-78200126 on Build GRCh38
rs143676391 Chr.11: 78200129-78200129 on Build GRCh38
rs967464879 Chr.11: 78200269-78200269 on Build GRCh38
rs776334056 Chr.11: 78200284-78200285 on Build GRCh38
rs745355873 Chr.11: 78200165-78200165 on Build GRCh38
rs752491296 Chr.11: 78200141-78200141 on Build GRCh38
rs752898893 Chr.11: 78200213-78200213 on Build GRCh38
rs946821655 Chr.11: 78200299-78200299 on Build GRCh38
rs1027408533 Chr.11: 78200199-78200199 on Build GRCh38
rs763121499 Chr.11: 78200279-78200279 on Build GRCh38
rs762637041 Chr.11: 78200293-78200293 on Build GRCh38
rs775599351 Chr.11: 78200273-78200273 on Build GRCh38
rs773747750 Chr.11: 78200256-78200256 on Build GRCh38
rs1010990616 Chr.11: 78200154-78200154 on Build GRCh38
rs200421480 Chr.11: 78200194-78200194 on Build GRCh38
rs758143269 Chr.11: 78200214-78200214 on Build GRCh38
rs933574184 Chr.11: 78200294-78200294 on Build GRCh38
rs2101310 Chr.11: 78200288-78200288 on Build GRCh38
rs375190563 Chr.11: 78200127-78200127 on Build GRCh38
rs938196957 Chr.11: 78200261-78200261 on Build GRCh38
rs548497676 Chr.11: 78200237-78200237 on Build GRCh38
rs902538822 Chr.11: 78200306-78200306 on Build GRCh38
rs771286034 Chr.11: 78200150-78200152 on Build GRCh38
rs747354109 Chr.11: 78200265-78200265 on Build GRCh38
rs200565562 Chr.11: 78200150-78200150 on Build GRCh38
rs759646870 Chr.11: 78200196-78200196 on Build GRCh38
rs115848453 Chr.11: 78200178-78200178 on Build GRCh38
rs369905425 Chr.11: 78200189-78200189 on Build GRCh38
rs949677547 Chr.11: 78200251-78200251 on Build GRCh38
rs796438637 Chr.11: 78200317-78200317 on Build GRCh38
rs775004717 Chr.11: 78200242-78200266 on Build GRCh38
rs928169131 Chr.11: 78200259-78200259 on Build GRCh38
rs746892951 Chr.11: 78200159-78200159 on Build GRCh38
rs772591751 Chr.11: 78200249-78200249 on Build GRCh38

View Full Product Details