Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101720018-101720275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHPT1
Entrez Gene ID
56994
Gene Name
choline phosphotransferase 1
Gene Aliases
CPT, CPT1
UniGene
Hs.293077
Gene Chromosome Location
Chr.12: 101697639-101752038 on Build GRCh38
Transcripts
NM_020244.2 XM_011538574.1 XM_011538575.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6628578 c.662A>T p.E221V Chr.12: 101720136-101720136 on Build GRCh38
3954058 c.658C>A p.L220I Chr.12: 101720132-101720132 on Build GRCh38
4672220 c.780+2T>C p.? Chr.12: 101720256-101720256 on Build GRCh38
3810792 c.677T>G p.I226S Chr.12: 101720151-101720151 on Build GRCh38
5933012 c.742C>T p.L248F Chr.12: 101720216-101720216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772552419 Chr.12: 101720202-101720202 on Build GRCh38
rs745362952 Chr.12: 101720094-101720097 on Build GRCh38
rs144773874 Chr.12: 101720150-101720150 on Build GRCh38
rs866519640 Chr.12: 101720244-101720244 on Build GRCh38
rs1051129023 Chr.12: 101720156-101720156 on Build GRCh38
rs931274647 Chr.12: 101720195-101720195 on Build GRCh38
rs766046295 Chr.12: 101720072-101720072 on Build GRCh38
rs563217696 Chr.12: 101720053-101720053 on Build GRCh38
rs370580831 Chr.12: 101720228-101720228 on Build GRCh38
rs560783909 Chr.12: 101720132-101720132 on Build GRCh38
rs758375136 Chr.12: 101720098-101720101 on Build GRCh38
rs765946077 Chr.12: 101720226-101720226 on Build GRCh38
rs1048688009 Chr.12: 101720229-101720229 on Build GRCh38
rs754768937 Chr.12: 101720085-101720085 on Build GRCh38
rs531942117 Chr.12: 101720067-101720067 on Build GRCh38
rs953925568 Chr.12: 101720139-101720139 on Build GRCh38
rs762467239 Chr.12: 101720205-101720205 on Build GRCh38
rs140053199 Chr.12: 101720157-101720157 on Build GRCh38
rs772370561 Chr.12: 101720123-101720123 on Build GRCh38
rs538281829 Chr.12: 101720166-101720166 on Build GRCh38
rs925670173 Chr.12: 101720077-101720077 on Build GRCh38
rs780002109 Chr.12: 101720101-101720104 on Build GRCh38
rs913487340 Chr.12: 101720111-101720111 on Build GRCh38
rs768959982 Chr.12: 101720153-101720153 on Build GRCh38
rs751301653 Chr.12: 101720078-101720078 on Build GRCh38
rs377678362 Chr.12: 101720164-101720164 on Build GRCh38
rs757335545 Chr.12: 101720115-101720115 on Build GRCh38
rs748091326 Chr.12: 101720107-101720107 on Build GRCh38
rs781180279 Chr.12: 101720086-101720086 on Build GRCh38
rs985209811 Chr.12: 101720144-101720144 on Build GRCh38
rs979863725 Chr.12: 101720052-101720052 on Build GRCh38
rs201833858 Chr.12: 101720116-101720116 on Build GRCh38
rs780436412 Chr.12: 101720137-101720137 on Build GRCh38

View Full Product Details