Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48604611-48604884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR13
Entrez Gene ID
64743
Gene Name
WD repeat domain 13
Gene Aliases
MG21
UniGene
Hs.521973
Gene Chromosome Location
Chr.X: 48597492-48605194 on Build GRCh38
Transcripts
NM_001166426.1 NM_017883.4 NR_029427.1 XM_006724539.1 XM_006724541.3 XM_011543947.2

SNP Detail

SNP ID Chromosome Location
rs782370120 Chr.X: 48604803-48604803 on Build GRCh38
rs782775494 Chr.X: 48604842-48604842 on Build GRCh38
rs782046823 Chr.X: 48604823-48604823 on Build GRCh38
rs781951300 Chr.X: 48604832-48604832 on Build GRCh38
rs144018865 Chr.X: 48604852-48604852 on Build GRCh38
rs782033540 Chr.X: 48604729-48604729 on Build GRCh38
rs782130859 Chr.X: 48604706-48604707 on Build GRCh38
rs782704022 Chr.X: 48604827-48604827 on Build GRCh38
rs782255821 Chr.X: 48604813-48604813 on Build GRCh38
rs376915714 Chr.X: 48604814-48604814 on Build GRCh38
rs943311043 Chr.X: 48604785-48604785 on Build GRCh38
rs781989150 Chr.X: 48604808-48604808 on Build GRCh38
rs911828153 Chr.X: 48604776-48604776 on Build GRCh38
rs782095129 Chr.X: 48604841-48604841 on Build GRCh38
rs782031239 Chr.X: 48604821-48604821 on Build GRCh38
rs782211911 Chr.X: 48604801-48604801 on Build GRCh38
rs782794264 Chr.X: 48604719-48604719 on Build GRCh38
rs371664708 Chr.X: 48604636-48604636 on Build GRCh38
rs781940273 Chr.X: 48604753-48604753 on Build GRCh38
rs923929469 Chr.X: 48604654-48604654 on Build GRCh38
rs933986434 Chr.X: 48604670-48604670 on Build GRCh38

View Full Product Details