Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113113720-113113993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F7
Entrez Gene ID
2155
Gene Name
coagulation factor VII
Gene Aliases
SPCA
UniGene
Hs.36989
Gene Chromosome Location
Chr.13: 113105773-113120681 on Build GRCh38
Transcripts
NM_000131.4 NM_001267554.1 NM_019616.3 NR_051961.1 XM_006719963.3 XM_011537474.2 XM_011537475.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7165450 c.348G>C p.Q116H Chr.13: 113113878-113113878 on Build GRCh38
5786119 c.420C>A p.N140K Chr.13: 113113950-113113950 on Build GRCh38
1677814 c.319G>A p.G107R Chr.13: 113113849-113113849 on Build GRCh38
5920590 c.193C>T p.P65S Chr.13: 113113930-113113930 on Build GRCh38
3700384 c.316+3G>A p.? Chr.13: 113113779-113113779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056381677 Chr.13: 113113801-113113801 on Build GRCh38
rs200693831 Chr.13: 113113959-113113959 on Build GRCh38
rs141678243 Chr.13: 113113939-113113939 on Build GRCh38
rs367552672 Chr.13: 113113872-113113872 on Build GRCh38
rs777385390 Chr.13: 113113849-113113849 on Build GRCh38
rs767843171 Chr.13: 113113777-113113777 on Build GRCh38
rs374056563 Chr.13: 113113808-113113808 on Build GRCh38
rs780805759 Chr.13: 113113814-113113814 on Build GRCh38
rs774511988 Chr.13: 113113772-113113772 on Build GRCh38
rs762761780 Chr.13: 113113739-113113739 on Build GRCh38
rs776796178 Chr.13: 113113945-113113945 on Build GRCh38
rs374825658 Chr.13: 113113783-113113783 on Build GRCh38
rs1009554951 Chr.13: 113113822-113113822 on Build GRCh38
rs745925935 Chr.13: 113113911-113113911 on Build GRCh38
rs747937888 Chr.13: 113113829-113113829 on Build GRCh38
rs771154432 Chr.13: 113113746-113113746 on Build GRCh38
rs576456456 Chr.13: 113113914-113113914 on Build GRCh38
rs765720786 Chr.13: 113113950-113113950 on Build GRCh38
rs755667709 Chr.13: 113113966-113113966 on Build GRCh38
rs777626344 Chr.13: 113113967-113113967 on Build GRCh38
rs757650344 Chr.13: 113113807-113113807 on Build GRCh38
rs761077461 Chr.13: 113113785-113113785 on Build GRCh38
rs121964932 Chr.13: 113113879-113113879 on Build GRCh38
rs1056071555 Chr.13: 113113961-113113961 on Build GRCh38
rs121964933 Chr.13: 113113893-113113893 on Build GRCh38
rs150525536 Chr.13: 113113946-113113946 on Build GRCh38
rs774636523 Chr.13: 113113894-113113894 on Build GRCh38
rs765874428 Chr.13: 113113902-113113902 on Build GRCh38
rs370535984 Chr.13: 113113774-113113774 on Build GRCh38
rs181538903 Chr.13: 113113960-113113960 on Build GRCh38
rs754398895 Chr.13: 113113795-113113795 on Build GRCh38
rs751119841 Chr.13: 113113958-113113958 on Build GRCh38
rs755972259 Chr.13: 113113830-113113830 on Build GRCh38
rs770921472 Chr.13: 113113888-113113888 on Build GRCh38
rs954706365 Chr.13: 113113848-113113848 on Build GRCh38
rs772471140 Chr.13: 113113912-113113912 on Build GRCh38
rs941222732 Chr.13: 113113742-113113742 on Build GRCh38
rs926423257 Chr.13: 113113754-113113754 on Build GRCh38
rs760766801 Chr.13: 113113929-113113929 on Build GRCh38
rs868060127 Chr.13: 113113852-113113852 on Build GRCh38
rs1042516833 Chr.13: 113113824-113113824 on Build GRCh38
rs764547586 Chr.13: 113113794-113113794 on Build GRCh38

View Full Product Details