Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 23566687-23566944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MKRN3
Entrez Gene ID
7681
Gene Name
makorin ring finger protein 3
Gene Aliases
CPPB2, D15S9, RNF63, ZFP127, ZNF127
UniGene
Hs.679587
Gene Chromosome Location
Chr.15: 23565307-23568020 on Build GRCh38
Transcripts
NM_005664.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4520075 c.1040G>A p.W347* Chr.15: 23566822-23566822 on Build GRCh38
392262 c.1083delC p.P362fs*34 Chr.15: 23566865-23566865 on Build GRCh38
959900 c.1053A>G p.R351R Chr.15: 23566835-23566835 on Build GRCh38
5916436 c.1007A>G p.N336S Chr.15: 23566789-23566789 on Build GRCh38
6450953 c.1072G>T p.V358F Chr.15: 23566854-23566854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760258851 Chr.15: 23566747-23566747 on Build GRCh38
rs772854066 Chr.15: 23566732-23566732 on Build GRCh38
rs1033077110 Chr.15: 23566731-23566731 on Build GRCh38
rs927754218 Chr.15: 23566748-23566748 on Build GRCh38
rs370114150 Chr.15: 23566865-23566865 on Build GRCh38
rs769287405 Chr.15: 23566858-23566858 on Build GRCh38
rs780654896 Chr.15: 23566833-23566833 on Build GRCh38
rs150392001 Chr.15: 23566893-23566893 on Build GRCh38
rs748074227 Chr.15: 23566825-23566825 on Build GRCh38
rs376939836 Chr.15: 23566789-23566789 on Build GRCh38
rs373400101 Chr.15: 23566761-23566761 on Build GRCh38
rs148323961 Chr.15: 23566799-23566799 on Build GRCh38
rs142151788 Chr.15: 23566760-23566760 on Build GRCh38
rs776224446 Chr.15: 23566838-23566838 on Build GRCh38
rs375705600 Chr.15: 23566816-23566816 on Build GRCh38
rs776038669 Chr.15: 23566757-23566757 on Build GRCh38
rs146606388 Chr.15: 23566766-23566766 on Build GRCh38
rs780216730 Chr.15: 23566804-23566804 on Build GRCh38
rs112128357 Chr.15: 23566857-23566857 on Build GRCh38
rs752680337 Chr.15: 23566916-23566916 on Build GRCh38
rs770427898 Chr.15: 23566753-23566753 on Build GRCh38
rs552926693 Chr.15: 23566775-23566775 on Build GRCh38
rs1009533218 Chr.15: 23566878-23566878 on Build GRCh38
rs766456029 Chr.15: 23566892-23566892 on Build GRCh38
rs770448149 Chr.15: 23566835-23566835 on Build GRCh38
rs535727288 Chr.15: 23566879-23566879 on Build GRCh38
rs1010605011 Chr.15: 23566827-23566827 on Build GRCh38
rs762392432 Chr.15: 23566875-23566875 on Build GRCh38
rs753814036 Chr.15: 23566809-23566809 on Build GRCh38
rs771912869 Chr.15: 23566828-23566828 on Build GRCh38
rs201462867 Chr.15: 23566874-23566874 on Build GRCh38
rs765287140 Chr.15: 23566903-23566903 on Build GRCh38
rs768002621 Chr.15: 23566762-23566762 on Build GRCh38
rs138332709 Chr.15: 23566895-23566895 on Build GRCh38
rs748041651 Chr.15: 23566897-23566897 on Build GRCh38
rs140467331 Chr.15: 23566883-23566883 on Build GRCh38
rs879255240 Chr.15: 23566877-23566877 on Build GRCh38
rs370224529 Chr.15: 23566735-23566735 on Build GRCh38
rs373441581 Chr.15: 23566815-23566815 on Build GRCh38
rs760981395 Chr.15: 23566885-23566885 on Build GRCh38
rs535543560 Chr.15: 23566853-23566853 on Build GRCh38

View Full Product Details