Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141383036-141383265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHGA1
Entrez Gene ID
56114
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
PCDH-GAMMA-A1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141330685-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Gene Symbol
PCDHGA2
Entrez Gene ID
56113
Gene Name
protocadherin gamma subfamily A, 2
Gene Aliases
PCDH-GAMMA-A2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141338760-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Gene Symbol
PCDHGA3
Entrez Gene ID
56112
Gene Name
protocadherin gamma subfamily A, 3
Gene Aliases
PCDH-GAMMA-A3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141344034-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Gene Symbol
PCDHGA4
Entrez Gene ID
56111
Gene Name
protocadherin gamma subfamily A, 4
Gene Aliases
PCDH-GAMMA-A4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141355025-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Gene Symbol
PCDHGA5
Entrez Gene ID
56110
Gene Name
protocadherin gamma subfamily A, 5
Gene Aliases
CDH-GAMMA-A5, ME3, PCDH-GAMMA-A5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141364331-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Gene Symbol
PCDHGA6
Entrez Gene ID
56109
Gene Name
protocadherin gamma subfamily A, 6
Gene Aliases
PCDH-GAMMA-A6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141374084-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Gene Symbol
PCDHGA7
Entrez Gene ID
56108
Gene Name
protocadherin gamma subfamily A, 7
Gene Aliases
PCDH-GAMMA-A7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141382739-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Gene Symbol
PCDHGB1
Entrez Gene ID
56104
Gene Name
protocadherin gamma subfamily B, 1
Gene Aliases
PCDH-GAMMA-B1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141350261-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Gene Symbol
PCDHGB2
Entrez Gene ID
56103
Gene Name
protocadherin gamma subfamily B, 2
Gene Aliases
PCDH-GAMMA-B2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141360136-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Gene Symbol
PCDHGB3
Entrez Gene ID
56102
Gene Name
protocadherin gamma subfamily B, 3
Gene Aliases
PCDH-GAMMA-B3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141370264-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032087.2 NM_018922.2 NM_018923.2 NM_018924.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5455359 c.217C>T p.Q73* Chr.5: 141383116-141383116 on Build GRCh38
6740967 c.216G>A p.T72T Chr.5: 141383115-141383115 on Build GRCh38
6217148 c.282G>T p.E94D Chr.5: 141383181-141383181 on Build GRCh38
5466312 c.346C>A p.L116I Chr.5: 141383245-141383245 on Build GRCh38
4478087 c.220C>T p.L74F Chr.5: 141383119-141383119 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773972778 Chr.5: 141383085-141383085 on Build GRCh38
rs982604012 Chr.5: 141383071-141383072 on Build GRCh38
rs775727094 Chr.5: 141383175-141383175 on Build GRCh38
rs771918652 Chr.5: 141383101-141383102 on Build GRCh38
rs775672220 Chr.5: 141383105-141383105 on Build GRCh38
rs372087170 Chr.5: 141383204-141383204 on Build GRCh38
rs200500982 Chr.5: 141383152-141383152 on Build GRCh38
rs373805952 Chr.5: 141383086-141383086 on Build GRCh38
rs562167618 Chr.5: 141383109-141383109 on Build GRCh38
rs750687055 Chr.5: 141383137-141383137 on Build GRCh38
rs991434235 Chr.5: 141383102-141383102 on Build GRCh38
rs757654723 Chr.5: 141383125-141383125 on Build GRCh38
rs770794066 Chr.5: 141383079-141383079 on Build GRCh38
rs755782697 Chr.5: 141383203-141383203 on Build GRCh38
rs749274128 Chr.5: 141383154-141383154 on Build GRCh38
rs778761472 Chr.5: 141383222-141383222 on Build GRCh38
rs765730698 Chr.5: 141383195-141383195 on Build GRCh38
rs771829169 Chr.5: 141383174-141383174 on Build GRCh38
rs985561606 Chr.5: 141383185-141383185 on Build GRCh38
rs376511249 Chr.5: 141383068-141383068 on Build GRCh38
rs780853671 Chr.5: 141383065-141383065 on Build GRCh38
rs76873890 Chr.5: 141383223-141383223 on Build GRCh38
rs574670513 Chr.5: 141383199-141383199 on Build GRCh38
rs747790551 Chr.5: 141383073-141383073 on Build GRCh38
rs1037294449 Chr.5: 141383134-141383134 on Build GRCh38
rs546342817 Chr.5: 141383160-141383160 on Build GRCh38
rs781049830 Chr.5: 141383141-141383141 on Build GRCh38
rs367646444 Chr.5: 141383217-141383217 on Build GRCh38
rs769758778 Chr.5: 141383159-141383159 on Build GRCh38
rs765646397 Chr.5: 141383133-141383133 on Build GRCh38
rs745716373 Chr.5: 141383166-141383166 on Build GRCh38
rs538619604 Chr.5: 141383138-141383138 on Build GRCh38
rs760790560 Chr.5: 141383106-141383106 on Build GRCh38
rs1024955582 Chr.5: 141383126-141383126 on Build GRCh38
rs771882574 Chr.5: 141383226-141383226 on Build GRCh38
rs7704696 Chr.5: 141383196-141383196 on Build GRCh38
rs770719921 Chr.5: 141383160-141383160 on Build GRCh38
rs762386846 Chr.5: 141383188-141383188 on Build GRCh38
rs760844021 Chr.5: 141383186-141383186 on Build GRCh38
rs749152940 Chr.5: 141383078-141383078 on Build GRCh38
rs371358932 Chr.5: 141383143-141383143 on Build GRCh38
rs184479480 Chr.5: 141383197-141383197 on Build GRCh38
rs754398892 Chr.5: 141383110-141383110 on Build GRCh38
rs572411306 Chr.5: 141383187-141383187 on Build GRCh38
rs375026409 Chr.5: 141383215-141383215 on Build GRCh38
rs774337554 Chr.5: 141383161-141383161 on Build GRCh38
rs911488617 Chr.5: 141383193-141383193 on Build GRCh38
rs368180461 Chr.5: 141383201-141383201 on Build GRCh38
rs768364409 Chr.5: 141383056-141383056 on Build GRCh38
rs200016406 Chr.5: 141383092-141383092 on Build GRCh38
rs370612007 Chr.5: 141383069-141383069 on Build GRCh38

View Full Product Details