Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 71301466-71301723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OGFRL1
Entrez Gene ID
79627
Gene Name
opioid growth factor receptor like 1
Gene Aliases
dJ331H24.1
UniGene
Hs.648434
Gene Chromosome Location
Chr.6: 71288296-71309059 on Build GRCh38
Transcripts
NM_001324266.1 NM_024576.4 XM_011536117.2 XM_011536118.2 XM_017011303.1 XM_017011304.1 XM_017011305.1 XM_017011306.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1081325 c.978G>A p.Q326Q Chr.6: 71301671-71301671 on Build GRCh38
4512254 c.897C>T p.F299F Chr.6: 71301590-71301590 on Build GRCh38
451752 c.893G>A p.R298Q Chr.6: 71301586-71301586 on Build GRCh38
3161606 c.946C>T p.R316* Chr.6: 71301639-71301639 on Build GRCh38
3161607 c.960G>A p.S320S Chr.6: 71301653-71301653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765501956 Chr.6: 71301593-71301593 on Build GRCh38
rs983446903 Chr.6: 71301586-71301586 on Build GRCh38
rs546446159 Chr.6: 71301521-71301521 on Build GRCh38
rs563435080 Chr.6: 71301549-71301549 on Build GRCh38
rs140095481 Chr.6: 71301615-71301615 on Build GRCh38
rs770680242 Chr.6: 71301633-71301633 on Build GRCh38
rs149936579 Chr.6: 71301557-71301557 on Build GRCh38
rs764093237 Chr.6: 71301585-71301585 on Build GRCh38
rs111886368 Chr.6: 71301691-71301691 on Build GRCh38
rs764730106 Chr.6: 71301650-71301650 on Build GRCh38
rs567112388 Chr.6: 71301602-71301602 on Build GRCh38
rs768106483 Chr.6: 71301560-71301560 on Build GRCh38
rs764248202 Chr.6: 71301562-71301563 on Build GRCh38
rs201295197 Chr.6: 71301649-71301649 on Build GRCh38
rs770681585 Chr.6: 71301550-71301550 on Build GRCh38
rs143683047 Chr.6: 71301653-71301653 on Build GRCh38
rs774182663 Chr.6: 71301635-71301635 on Build GRCh38
rs1045195751 Chr.6: 71301524-71301524 on Build GRCh38
rs751334575 Chr.6: 71301598-71301598 on Build GRCh38
rs199584243 Chr.6: 71301590-71301590 on Build GRCh38
rs752616694 Chr.6: 71301672-71301672 on Build GRCh38
rs1049085373 Chr.6: 71301614-71301614 on Build GRCh38
rs757418904 Chr.6: 71301591-71301591 on Build GRCh38
rs551206380 Chr.6: 71301509-71301509 on Build GRCh38
rs772587548 Chr.6: 71301640-71301640 on Build GRCh38
rs371570884 Chr.6: 71301552-71301552 on Build GRCh38
rs748883572 Chr.6: 71301631-71301631 on Build GRCh38
rs150212567 Chr.6: 71301669-71301669 on Build GRCh38
rs540548321 Chr.6: 71301682-71301682 on Build GRCh38
rs996529829 Chr.6: 71301687-71301687 on Build GRCh38
rs765415623 Chr.6: 71301658-71301658 on Build GRCh38
rs772874288 Chr.6: 71301539-71301539 on Build GRCh38
rs755595849 Chr.6: 71301608-71301608 on Build GRCh38
rs146479750 Chr.6: 71301578-71301578 on Build GRCh38
rs776561377 Chr.6: 71301652-71301652 on Build GRCh38
rs776080362 Chr.6: 71301648-71301648 on Build GRCh38
rs754723890 Chr.6: 71301663-71301663 on Build GRCh38
rs755989224 Chr.6: 71301689-71301689 on Build GRCh38
rs373599406 Chr.6: 71301607-71301607 on Build GRCh38
rs746194897 Chr.6: 71301639-71301639 on Build GRCh38
rs750743575 Chr.6: 71301510-71301510 on Build GRCh38
rs777810665 Chr.6: 71301690-71301690 on Build GRCh38
rs769223481 Chr.6: 71301517-71301517 on Build GRCh38
rs750678168 Chr.6: 71301661-71301661 on Build GRCh38

View Full Product Details