Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 146156326-146156067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOTCH2NL
Entrez Gene ID
388677
Gene Name
notch 2 N-terminal like
Gene Aliases
N2N
UniGene
Hs.655156
Gene Chromosome Location
Chr.1: 146151908-146229032 on Build GRCh38
Transcripts
NM_203458.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
276369 c.694G>T p.E232* Chr.1: 146156170-146156170 on Build GRCh38
3704925 c.711G>T p.*237Y Chr.1: 146156153-146156153 on Build GRCh38
674826 c.711G>C p.*237Y Chr.1: 146156153-146156153 on Build GRCh38
2231883 c.669_672delAGAC p.D224fs*>12 Chr.1: 146156192-146156195 on Build GRCh38
1294920 c.700G>A p.D234N Chr.1: 146156164-146156164 on Build GRCh38
View More

View Full Product Details