Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 141919672-141919905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR9A4
Entrez Gene ID
130075
Gene Name
olfactory receptor family 9 subfamily A member 4
Gene Aliases
-
UniGene
Hs.690171
Gene Chromosome Location
Chr.7: 141918876-141919820 on Build GRCh38
Transcripts
NM_001001656.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
173013 c.919C>T p.R307C Chr.7: 141919794-141919794 on Build GRCh38
1722431 c.852C>T p.F284F Chr.7: 141919727-141919727 on Build GRCh38
6299264 c.865A>T p.I289F Chr.7: 141919740-141919740 on Build GRCh38
5588539 c.924C>T p.C308C Chr.7: 141919799-141919799 on Build GRCh38
3922849 c.867C>T p.I289I Chr.7: 141919742-141919742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782703595 Chr.7: 141919785-141919785 on Build GRCh38
rs201481115 Chr.7: 141919752-141919752 on Build GRCh38
rs782633602 Chr.7: 141919795-141919795 on Build GRCh38
rs782743378 Chr.7: 141919702-141919702 on Build GRCh38
rs781818850 Chr.7: 141919787-141919787 on Build GRCh38
rs1025033837 Chr.7: 141919748-141919748 on Build GRCh38
rs782069520 Chr.7: 141919758-141919758 on Build GRCh38
rs371951224 Chr.7: 141919759-141919759 on Build GRCh38
rs782374978 Chr.7: 141919729-141919729 on Build GRCh38
rs782232749 Chr.7: 141919855-141919855 on Build GRCh38
rs540557231 Chr.7: 141919794-141919794 on Build GRCh38
rs781956266 Chr.7: 141919842-141919842 on Build GRCh38
rs782285797 Chr.7: 141919859-141919859 on Build GRCh38
rs782172749 Chr.7: 141919721-141919721 on Build GRCh38
rs190975595 Chr.7: 141919833-141919833 on Build GRCh38
rs782178741 Chr.7: 141919828-141919828 on Build GRCh38
rs371819757 Chr.7: 141919705-141919705 on Build GRCh38
rs782598660 Chr.7: 141919723-141919723 on Build GRCh38
rs782663644 Chr.7: 141919843-141919843 on Build GRCh38
rs782005214 Chr.7: 141919744-141919744 on Build GRCh38
rs782125001 Chr.7: 141919768-141919768 on Build GRCh38
rs564898434 Chr.7: 141919822-141919822 on Build GRCh38
rs782539235 Chr.7: 141919821-141919821 on Build GRCh38
rs574844918 Chr.7: 141919728-141919728 on Build GRCh38
rs963581285 Chr.7: 141919780-141919780 on Build GRCh38
rs782263996 Chr.7: 141919796-141919796 on Build GRCh38
rs370705799 Chr.7: 141919698-141919698 on Build GRCh38
rs1052336420 Chr.7: 141919711-141919711 on Build GRCh38
rs573034609 Chr.7: 141919775-141919775 on Build GRCh38
rs782685161 Chr.7: 141919697-141919697 on Build GRCh38
rs781973881 Chr.7: 141919764-141919764 on Build GRCh38
rs782322628 Chr.7: 141919722-141919722 on Build GRCh38
rs782790089 Chr.7: 141919770-141919770 on Build GRCh38
rs782445685 Chr.7: 141919781-141919781 on Build GRCh38

View Full Product Details