Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2132992-2133225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF2
Entrez Gene ID
3481
Gene Name
insulin like growth factor 2
Gene Aliases
C11orf43, GRDF, IGF-II, PP9974
UniGene
Hs.272259
Gene Chromosome Location
Chr.11: 2129112-2149603 on Build GRCh38
Transcripts
NM_000612.5 NM_001007139.5 NM_001127598.2 NM_001291861.2 NM_001291862.2 NR_003512.3

Gene Symbol
INS-IGF2
Entrez Gene ID
723961
Gene Name
INS-IGF2 readthrough
Gene Aliases
INSIGF
UniGene
Hs.272259
Gene Chromosome Location
Chr.11: 2129112-2161209 on Build GRCh38
Transcripts
NM_000612.5 NM_001007139.5 NM_001127598.2 NM_001291861.2 NM_001291862.2 NR_003512.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4949349 c.338T>A p.F113Y Chr.11: 2133192-2133192 on Build GRCh38
5596860 c.683C>T p.P228L Chr.11: 2133015-2133015 on Build GRCh38
6971847 c.583G>T p.G195C Chr.11: 2133115-2133115 on Build GRCh38
4852080 c.498C>A p.P166P Chr.11: 2133032-2133032 on Build GRCh38
136808 c.415G>A p.G139S Chr.11: 2133115-2133115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs14367 Chr.11: 2133170-2133170 on Build GRCh38
rs1042993851 Chr.11: 2133178-2133178 on Build GRCh38
rs377316111 Chr.11: 2133031-2133031 on Build GRCh38
rs554804044 Chr.11: 2133025-2133025 on Build GRCh38
rs200036157 Chr.11: 2133026-2133026 on Build GRCh38
rs150866176 Chr.11: 2133091-2133091 on Build GRCh38
rs747026832 Chr.11: 2133021-2133021 on Build GRCh38
rs1032405005 Chr.11: 2133051-2133051 on Build GRCh38
rs758849172 Chr.11: 2133070-2133070 on Build GRCh38
rs199646294 Chr.11: 2133041-2133041 on Build GRCh38
rs11545014 Chr.11: 2133013-2133013 on Build GRCh38
rs747266091 Chr.11: 2133079-2133079 on Build GRCh38
rs760894162 Chr.11: 2133082-2133082 on Build GRCh38
rs902646764 Chr.11: 2133014-2133014 on Build GRCh38
rs372036269 Chr.11: 2133104-2133104 on Build GRCh38
rs767815580 Chr.11: 2133032-2133032 on Build GRCh38
rs755482890 Chr.11: 2133061-2133061 on Build GRCh38
rs750321513 Chr.11: 2133203-2133203 on Build GRCh38
rs530101369 Chr.11: 2133133-2133133 on Build GRCh38
rs763681027 Chr.11: 2133035-2133035 on Build GRCh38
rs773624887 Chr.11: 2133018-2133018 on Build GRCh38
rs748459239 Chr.11: 2133058-2133058 on Build GRCh38
rs745901132 Chr.11: 2133198-2133198 on Build GRCh38
rs369452652 Chr.11: 2133114-2133114 on Build GRCh38
rs368814253 Chr.11: 2133087-2133087 on Build GRCh38
rs139194127 Chr.11: 2133089-2133089 on Build GRCh38
rs1006709496 Chr.11: 2133106-2133106 on Build GRCh38
rs773939360 Chr.11: 2133092-2133092 on Build GRCh38
rs781325329 Chr.11: 2133157-2133157 on Build GRCh38
rs757547834 Chr.11: 2133156-2133156 on Build GRCh38
rs771327940 Chr.11: 2133083-2133083 on Build GRCh38
rs749953596 Chr.11: 2133058-2133058 on Build GRCh38
rs755455183 Chr.11: 2133011-2133012 on Build GRCh38
rs781299823 Chr.11: 2133086-2133086 on Build GRCh38
rs778075715 Chr.11: 2133076-2133076 on Build GRCh38
rs61732764 Chr.11: 2133063-2133063 on Build GRCh38
rs765719749 Chr.11: 2133110-2133110 on Build GRCh38
rs371682005 Chr.11: 2133118-2133118 on Build GRCh38
rs771138084 Chr.11: 2133022-2133022 on Build GRCh38
rs146962483 Chr.11: 2133069-2133069 on Build GRCh38
rs927034247 Chr.11: 2133115-2133115 on Build GRCh38
rs200836416 Chr.11: 2133103-2133103 on Build GRCh38
rs779532835 Chr.11: 2133049-2133063 on Build GRCh38
rs761436263 Chr.11: 2133100-2133100 on Build GRCh38
rs778304668 Chr.11: 2133123-2133123 on Build GRCh38
rs138399775 Chr.11: 2133020-2133020 on Build GRCh38
rs767032426 Chr.11: 2133056-2133056 on Build GRCh38
rs980331479 Chr.11: 2133068-2133068 on Build GRCh38
rs1050342 Chr.11: 2133012-2133012 on Build GRCh38
rs868243135 Chr.11: 2133174-2133174 on Build GRCh38
rs376878619 Chr.11: 2133202-2133202 on Build GRCh38
rs1065443 Chr.11: 2133149-2133149 on Build GRCh38
rs756720073 Chr.11: 2133050-2133050 on Build GRCh38
rs769766130 Chr.11: 2133201-2133201 on Build GRCh38
rs375129654 Chr.11: 2133109-2133109 on Build GRCh38

View Full Product Details