Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33413880-33414139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHF1
Entrez Gene ID
5252
Gene Name
PHD finger protein 1
Gene Aliases
MTF2L2, PCL1, PHF2, TDRD19C, hPHF1
UniGene
Hs.166204
Gene Chromosome Location
Chr.6: 33410751-33416453 on Build GRCh38
Transcripts
NM_002636.4 NM_024165.2 NR_027692.1 XM_011514662.1 XM_011514663.1 XM_011514664.1 XM_011514665.1 XM_011514666.1 XM_011514669.1 XM_011514670.2 XM_017010939.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6744903 c.712C>T p.R238C Chr.6: 33414069-33414069 on Build GRCh38
3349899 c.748C>T p.R250C Chr.6: 33414105-33414105 on Build GRCh38
4648988 c.749G>A p.R250H Chr.6: 33414106-33414106 on Build GRCh38
3349898 c.734G>A p.R245Q Chr.6: 33414091-33414091 on Build GRCh38
4614352 c.715delG p.G240fs*137 Chr.6: 33414072-33414072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558515434 Chr.6: 33413933-33413933 on Build GRCh38
rs757256695 Chr.6: 33414091-33414091 on Build GRCh38
rs780780759 Chr.6: 33414060-33414061 on Build GRCh38
rs963901269 Chr.6: 33413939-33413939 on Build GRCh38
rs144032908 Chr.6: 33414069-33414069 on Build GRCh38
rs750111165 Chr.6: 33414011-33414012 on Build GRCh38
rs761582706 Chr.6: 33413959-33413959 on Build GRCh38
rs753892070 Chr.6: 33414071-33414071 on Build GRCh38
rs967061640 Chr.6: 33413918-33413919 on Build GRCh38
rs766728612 Chr.6: 33414095-33414095 on Build GRCh38
rs368875361 Chr.6: 33413956-33413956 on Build GRCh38
rs994237741 Chr.6: 33414098-33414098 on Build GRCh38
rs111685107 Chr.6: 33414111-33414111 on Build GRCh38
rs1013986664 Chr.6: 33413903-33413903 on Build GRCh38
rs1038069938 Chr.6: 33414089-33414089 on Build GRCh38
rs112930472 Chr.6: 33414112-33414112 on Build GRCh38
rs372535149 Chr.6: 33414050-33414050 on Build GRCh38
rs901263884 Chr.6: 33414061-33414061 on Build GRCh38
rs780599482 Chr.6: 33414006-33414006 on Build GRCh38
rs772709521 Chr.6: 33414035-33414035 on Build GRCh38
rs769087516 Chr.6: 33414017-33414017 on Build GRCh38
rs746672037 Chr.6: 33414007-33414010 on Build GRCh38
rs749793990 Chr.6: 33414009-33414009 on Build GRCh38
rs989058311 Chr.6: 33413915-33413915 on Build GRCh38
rs767831440 Chr.6: 33414105-33414105 on Build GRCh38
rs761180131 Chr.6: 33414024-33414024 on Build GRCh38
rs377166111 Chr.6: 33414003-33414003 on Build GRCh38
rs369609725 Chr.6: 33414022-33414022 on Build GRCh38
rs765206511 Chr.6: 33414074-33414074 on Build GRCh38
rs759548466 Chr.6: 33414072-33414072 on Build GRCh38
rs978796580 Chr.6: 33413923-33413923 on Build GRCh38
rs913034832 Chr.6: 33413919-33413919 on Build GRCh38
rs751708549 Chr.6: 33414085-33414085 on Build GRCh38
rs537481871 Chr.6: 33413931-33413931 on Build GRCh38
rs911130972 Chr.6: 33413962-33413962 on Build GRCh38
rs778061123 Chr.6: 33414102-33414102 on Build GRCh38
rs554374549 Chr.6: 33413904-33413904 on Build GRCh38
rs577558781 Chr.6: 33413941-33413941 on Build GRCh38
rs750600236 Chr.6: 33414107-33414107 on Build GRCh38
rs771789201 Chr.6: 33414028-33414028 on Build GRCh38
rs1040984913 Chr.6: 33414029-33414029 on Build GRCh38

View Full Product Details