Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48610822-48611047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXB7
Entrez Gene ID
3217
Gene Name
homeobox B7
Gene Aliases
HHO.C1, HOX2, HOX2C, Hox-2.3
UniGene
Hs.436181
Gene Chromosome Location
Chr.17: 48607232-48611021 on Build GRCh38
Transcripts
NM_004502.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7095179 c.71G>A p.G24E Chr.17: 48610848-48610848 on Build GRCh38
6037968 c.19G>A p.A7T Chr.17: 48610900-48610900 on Build GRCh38
707057 c.92C>T p.S31F Chr.17: 48610827-48610827 on Build GRCh38
161593 c.21G>C p.A7A Chr.17: 48610898-48610898 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs988882930 Chr.17: 48610939-48610939 on Build GRCh38
rs771288533 Chr.17: 48610862-48610862 on Build GRCh38
rs368160113 Chr.17: 48610873-48610873 on Build GRCh38
rs747503560 Chr.17: 48610959-48610959 on Build GRCh38
rs41307615 Chr.17: 48611026-48611026 on Build GRCh38
rs753272394 Chr.17: 48610950-48610950 on Build GRCh38
rs139369803 Chr.17: 48610863-48610863 on Build GRCh38
rs142383237 Chr.17: 48611021-48611021 on Build GRCh38
rs754452042 Chr.17: 48610952-48610952 on Build GRCh38
rs750120989 Chr.17: 48610943-48610943 on Build GRCh38
rs930977725 Chr.17: 48610975-48610975 on Build GRCh38
rs149621604 Chr.17: 48610869-48610869 on Build GRCh38
rs146350891 Chr.17: 48610856-48610856 on Build GRCh38
rs16942475 Chr.17: 48611009-48611009 on Build GRCh38
rs972860470 Chr.17: 48610968-48610968 on Build GRCh38
rs767249080 Chr.17: 48610941-48610941 on Build GRCh38
rs541593072 Chr.17: 48611029-48611029 on Build GRCh38
rs781667080 Chr.17: 48610962-48610962 on Build GRCh38
rs751449761 Chr.17: 48610909-48610909 on Build GRCh38
rs912900892 Chr.17: 48610983-48610983 on Build GRCh38
rs569262374 Chr.17: 48610979-48610979 on Build GRCh38
rs761541738 Chr.17: 48610928-48610928 on Build GRCh38
rs974782744 Chr.17: 48610871-48610871 on Build GRCh38
rs548696991 Chr.17: 48611020-48611020 on Build GRCh38
rs1029242526 Chr.17: 48610889-48610889 on Build GRCh38
rs371705293 Chr.17: 48610947-48610947 on Build GRCh38
rs763954258 Chr.17: 48610901-48610901 on Build GRCh38
rs987635838 Chr.17: 48610937-48610937 on Build GRCh38
rs866068658 Chr.17: 48610872-48610872 on Build GRCh38
rs902311076 Chr.17: 48611010-48611010 on Build GRCh38
rs538394090 Chr.17: 48611008-48611008 on Build GRCh38
rs1036002040 Chr.17: 48610887-48610887 on Build GRCh38
rs755728332 Chr.17: 48610949-48610949 on Build GRCh38
rs7406910 Chr.17: 48610894-48610894 on Build GRCh38
rs1054753252 Chr.17: 48610993-48610994 on Build GRCh38
rs763009085 Chr.17: 48610899-48610899 on Build GRCh38
rs771471268 Chr.17: 48610960-48610960 on Build GRCh38
rs556138860 Chr.17: 48610902-48610902 on Build GRCh38
rs778176669 Chr.17: 48610953-48610953 on Build GRCh38

View Full Product Details