Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 27435750-27436007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTIF3
Entrez Gene ID
219402
Gene Name
mitochondrial translational initiation factor 3
Gene Aliases
IF3mt
UniGene
Hs.534582
Gene Chromosome Location
Chr.13: 27435639-27450900 on Build GRCh38
Transcripts
NM_001166261.1 NM_001166262.1 NM_001166263.1 NM_152912.4 XM_006719771.3 XM_006719772.3 XM_011534957.2 XM_011534958.2 XM_011534959.2 XM_011534960.2 XM_011534961.2 XM_011534962.2 XM_011534963.2 XM_017020417.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5990640 c.728T>G p.F243C Chr.13: 27435784-27435784 on Build GRCh38
4415909 c.729C>G p.F243L Chr.13: 27435783-27435783 on Build GRCh38
4703579 c.725C>T p.A242V Chr.13: 27435787-27435787 on Build GRCh38
6362522 c.688G>A p.V230I Chr.13: 27435824-27435824 on Build GRCh38
7148845 c.640C>T p.L214F Chr.13: 27435872-27435872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764011201 Chr.13: 27435902-27435902 on Build GRCh38
rs751175666 Chr.13: 27435904-27435904 on Build GRCh38
rs761423029 Chr.13: 27435909-27435909 on Build GRCh38
rs750936535 Chr.13: 27435882-27435883 on Build GRCh38
rs963346825 Chr.13: 27435900-27435900 on Build GRCh38
rs998669242 Chr.13: 27435792-27435792 on Build GRCh38
rs368296042 Chr.13: 27435944-27435944 on Build GRCh38
rs995804507 Chr.13: 27435903-27435903 on Build GRCh38
rs780590458 Chr.13: 27435799-27435799 on Build GRCh38
rs1027176195 Chr.13: 27435913-27435913 on Build GRCh38
rs567392549 Chr.13: 27435893-27435893 on Build GRCh38
rs1218825 Chr.13: 27435783-27435783 on Build GRCh38
rs773201911 Chr.13: 27435871-27435871 on Build GRCh38
rs748336585 Chr.13: 27435856-27435856 on Build GRCh38
rs750921639 Chr.13: 27435802-27435802 on Build GRCh38
rs141983211 Chr.13: 27435854-27435854 on Build GRCh38
rs758412376 Chr.13: 27435888-27435890 on Build GRCh38
rs756621481 Chr.13: 27435809-27435809 on Build GRCh38
rs375822703 Chr.13: 27435790-27435790 on Build GRCh38
rs762191462 Chr.13: 27435791-27435793 on Build GRCh38
rs916587417 Chr.13: 27435851-27435851 on Build GRCh38
rs148885989 Chr.13: 27435791-27435791 on Build GRCh38
rs780395213 Chr.13: 27435819-27435819 on Build GRCh38
rs982201023 Chr.13: 27435934-27435934 on Build GRCh38
rs765696159 Chr.13: 27435877-27435899 on Build GRCh38
rs1885989 Chr.13: 27435980-27435980 on Build GRCh38
rs940504265 Chr.13: 27435969-27435969 on Build GRCh38
rs907671864 Chr.13: 27435953-27435953 on Build GRCh38
rs191729623 Chr.13: 27435884-27435884 on Build GRCh38
rs201690059 Chr.13: 27435826-27435826 on Build GRCh38
rs767023553 Chr.13: 27435935-27435935 on Build GRCh38
rs536356900 Chr.13: 27435978-27435978 on Build GRCh38
rs756750858 Chr.13: 27435793-27435793 on Build GRCh38
rs749714164 Chr.13: 27435825-27435825 on Build GRCh38
rs775581370 Chr.13: 27435888-27435888 on Build GRCh38
rs769908353 Chr.13: 27435887-27435887 on Build GRCh38
rs772165698 Chr.13: 27435867-27435867 on Build GRCh38
rs775462597 Chr.13: 27435971-27435971 on Build GRCh38

View Full Product Details