Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 76833424-76833661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F2RL1
Entrez Gene ID
2150
Gene Name
F2R like trypsin receptor 1
Gene Aliases
GPR11, PAR2
UniGene
Hs.744181
Gene Chromosome Location
Chr.5: 76819008-76835315 on Build GRCh38
Transcripts
XM_017009223.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4804235 c.1031A>G p.Y344C Chr.5: 76833638-76833638 on Build GRCh38
4788150 c.846G>A p.R282R Chr.5: 76833453-76833453 on Build GRCh38
6587515 c.1043C>A p.S348* Chr.5: 76833650-76833650 on Build GRCh38
1070251 c.840G>T p.K280N Chr.5: 76833447-76833447 on Build GRCh38
4894085 c.961C>T p.H321Y Chr.5: 76833568-76833568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs917529421 Chr.5: 76833453-76833453 on Build GRCh38
rs187106934 Chr.5: 76833452-76833452 on Build GRCh38
rs955691308 Chr.5: 76833459-76833459 on Build GRCh38
rs765886275 Chr.5: 76833469-76833469 on Build GRCh38
rs747922576 Chr.5: 76833572-76833572 on Build GRCh38
rs764534865 Chr.5: 76833629-76833629 on Build GRCh38
rs779439389 Chr.5: 76833520-76833522 on Build GRCh38
rs760097382 Chr.5: 76833450-76833450 on Build GRCh38
rs764220227 Chr.5: 76833498-76833498 on Build GRCh38
rs1001582364 Chr.5: 76833569-76833569 on Build GRCh38
rs776250593 Chr.5: 76833600-76833600 on Build GRCh38
rs770414077 Chr.5: 76833622-76833622 on Build GRCh38
rs1025809601 Chr.5: 76833616-76833616 on Build GRCh38
rs755911067 Chr.5: 76833560-76833560 on Build GRCh38
rs781358902 Chr.5: 76833531-76833531 on Build GRCh38
rs751814011 Chr.5: 76833521-76833521 on Build GRCh38
rs905528105 Chr.5: 76833544-76833544 on Build GRCh38
rs370713776 Chr.5: 76833466-76833466 on Build GRCh38
rs770296967 Chr.5: 76833593-76833593 on Build GRCh38
rs763346141 Chr.5: 76833627-76833627 on Build GRCh38
rs2243083 Chr.5: 76833478-76833478 on Build GRCh38
rs914273365 Chr.5: 76833571-76833571 on Build GRCh38
rs144257769 Chr.5: 76833624-76833624 on Build GRCh38
rs267600695 Chr.5: 76833552-76833552 on Build GRCh38
rs753739168 Chr.5: 76833534-76833534 on Build GRCh38
rs754875227 Chr.5: 76833536-76833536 on Build GRCh38
rs778867086 Chr.5: 76833555-76833555 on Build GRCh38
rs757229771 Chr.5: 76833523-76833523 on Build GRCh38
rs745419972 Chr.5: 76833618-76833618 on Build GRCh38
rs777222086 Chr.5: 76833588-76833588 on Build GRCh38
rs771965396 Chr.5: 76833574-76833574 on Build GRCh38
rs376163464 Chr.5: 76833484-76833484 on Build GRCh38

View Full Product Details