Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 2847188-2847417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMILIN2
Entrez Gene ID
84034
Gene Name
elastin microfibril interfacer 2
Gene Aliases
EMILIN-2, FOAP-10
UniGene
Hs.532815
Gene Chromosome Location
Chr.18: 2847030-2914092 on Build GRCh38
Transcripts
NM_032048.2 XM_017026038.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5790815 c.117G>T p.R39R Chr.18: 2847305-2847305 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs572220705 Chr.18: 2847228-2847228 on Build GRCh38
rs542406908 Chr.18: 2847224-2847236 on Build GRCh38
rs112881773 Chr.18: 2847363-2847363 on Build GRCh38
rs1020040527 Chr.18: 2847384-2847384 on Build GRCh38
rs983625087 Chr.18: 2847333-2847333 on Build GRCh38
rs781538463 Chr.18: 2847307-2847307 on Build GRCh38
rs1049034768 Chr.18: 2847360-2847360 on Build GRCh38
rs900856093 Chr.18: 2847382-2847382 on Build GRCh38
rs756256350 Chr.18: 2847364-2847364 on Build GRCh38
rs909338699 Chr.18: 2847361-2847361 on Build GRCh38
rs974815680 Chr.18: 2847369-2847369 on Build GRCh38
rs1009867078 Chr.18: 2847383-2847383 on Build GRCh38
rs555490143 Chr.18: 2847205-2847205 on Build GRCh38
rs780508060 Chr.18: 2847378-2847378 on Build GRCh38
rs969744729 Chr.18: 2847236-2847236 on Build GRCh38
rs933317309 Chr.18: 2847375-2847375 on Build GRCh38
rs1052887873 Chr.18: 2847397-2847397 on Build GRCh38
rs751360407 Chr.18: 2847222-2847222 on Build GRCh38
rs951011785 Chr.18: 2847323-2847323 on Build GRCh38
rs749336005 Chr.18: 2847371-2847371 on Build GRCh38
rs534278634 Chr.18: 2847319-2847319 on Build GRCh38
rs114273128 Chr.18: 2847370-2847370 on Build GRCh38
rs868593440 Chr.18: 2847318-2847318 on Build GRCh38
rs1041287932 Chr.18: 2847380-2847380 on Build GRCh38
rs1035930067 Chr.18: 2847312-2847312 on Build GRCh38

View Full Product Details