Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6600702-6600927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRP8
Entrez Gene ID
23378
Gene Name
ribosomal RNA processing 8, methyltransferase, homolog (yeast)
Gene Aliases
KIAA0409, NML
UniGene
Hs.652255
Gene Chromosome Location
Chr.11: 6514830-6603678 on Build GRCh38
Transcripts
NM_015324.3 XM_011519955.2 XM_017017423.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
689901 c.1103G>A p.G368E Chr.11: 6600720-6600720 on Build GRCh38
5980039 c.1061A>G p.D354G Chr.11: 6600762-6600762 on Build GRCh38
1703513 c.1049T>C p.V350A Chr.11: 6600774-6600774 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs377756818 Chr.11: 6600807-6600807 on Build GRCh38
rs963228219 Chr.11: 6600731-6600731 on Build GRCh38
rs181419546 Chr.11: 6600789-6600789 on Build GRCh38
rs540425871 Chr.11: 6600846-6600846 on Build GRCh38
rs745760171 Chr.11: 6600795-6600795 on Build GRCh38
rs376752500 Chr.11: 6600740-6600740 on Build GRCh38
rs764391112 Chr.11: 6600822-6600822 on Build GRCh38
rs760862583 Chr.11: 6600814-6600814 on Build GRCh38
rs747763666 Chr.11: 6600762-6600762 on Build GRCh38
rs780699136 Chr.11: 6600748-6600748 on Build GRCh38
rs115960367 Chr.11: 6600781-6600781 on Build GRCh38
rs374978249 Chr.11: 6600733-6600735 on Build GRCh38
rs780072142 Chr.11: 6600829-6600829 on Build GRCh38
rs757889233 Chr.11: 6600734-6600734 on Build GRCh38
rs764454380 Chr.11: 6600814-6600816 on Build GRCh38
rs35552935 Chr.11: 6600849-6600850 on Build GRCh38
rs943257506 Chr.11: 6600894-6600894 on Build GRCh38
rs771706105 Chr.11: 6600843-6600843 on Build GRCh38
rs754473108 Chr.11: 6600742-6600742 on Build GRCh38
rs76589565 Chr.11: 6600882-6600882 on Build GRCh38
rs370232444 Chr.11: 6600881-6600881 on Build GRCh38
rs200917700 Chr.11: 6600811-6600811 on Build GRCh38
rs573140653 Chr.11: 6600893-6600893 on Build GRCh38
rs909338149 Chr.11: 6600867-6600867 on Build GRCh38
rs539386339 Chr.11: 6600777-6600777 on Build GRCh38
rs755761102 Chr.11: 6600768-6600768 on Build GRCh38
rs775633655 Chr.11: 6600812-6600812 on Build GRCh38
rs985281645 Chr.11: 6600815-6600815 on Build GRCh38
rs765652038 Chr.11: 6600883-6600883 on Build GRCh38
rs750903148 Chr.11: 6600741-6600741 on Build GRCh38
rs764823666 Chr.11: 6600775-6600775 on Build GRCh38

View Full Product Details