Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71475134-71475407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF19
Entrez Gene ID
7567
Gene Name
zinc finger protein 19
Gene Aliases
KOX12
UniGene
Hs.660076
Gene Chromosome Location
Chr.16: 71474073-71489351 on Build GRCh38
Transcripts
NM_006961.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
278246 c.1193G>T p.R398I Chr.16: 71475354-71475354 on Build GRCh38
6870878 c.1327G>T p.D443Y Chr.16: 71475220-71475220 on Build GRCh38
6053411 c.1353A>G p.E451E Chr.16: 71475194-71475194 on Build GRCh38
4883896 c.1242G>T p.E414D Chr.16: 71475305-71475305 on Build GRCh38
3511831 c.1219C>T p.H407Y Chr.16: 71475328-71475328 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775270031 Chr.16: 71475370-71475370 on Build GRCh38
rs992642240 Chr.16: 71475184-71475184 on Build GRCh38
rs756241913 Chr.16: 71475223-71475223 on Build GRCh38
rs777257301 Chr.16: 71475354-71475354 on Build GRCh38
rs752489954 Chr.16: 71475210-71475210 on Build GRCh38
rs374729640 Chr.16: 71475182-71475182 on Build GRCh38
rs371232458 Chr.16: 71475317-71475317 on Build GRCh38
rs201802139 Chr.16: 71475308-71475308 on Build GRCh38
rs756430996 Chr.16: 71475343-71475343 on Build GRCh38
rs566864799 Chr.16: 71475357-71475357 on Build GRCh38
rs754594966 Chr.16: 71475197-71475197 on Build GRCh38
rs546976870 Chr.16: 71475247-71475247 on Build GRCh38
rs544564621 Chr.16: 71475359-71475359 on Build GRCh38
rs768879984 Chr.16: 71475250-71475250 on Build GRCh38
rs542033526 Chr.16: 71475309-71475309 on Build GRCh38
rs750940515 Chr.16: 71475168-71475168 on Build GRCh38
rs748308166 Chr.16: 71475340-71475340 on Build GRCh38
rs762453399 Chr.16: 71475377-71475377 on Build GRCh38
rs749513750 Chr.16: 71475249-71475249 on Build GRCh38
rs968865777 Chr.16: 71475227-71475227 on Build GRCh38
rs368395546 Chr.16: 71475268-71475268 on Build GRCh38
rs765994712 Chr.16: 71475272-71475272 on Build GRCh38
rs751312104 Chr.16: 71475380-71475380 on Build GRCh38
rs141096366 Chr.16: 71475344-71475344 on Build GRCh38
rs753561375 Chr.16: 71475193-71475193 on Build GRCh38
rs777806223 Chr.16: 71475212-71475212 on Build GRCh38
rs758280396 Chr.16: 71475303-71475303 on Build GRCh38
rs924514915 Chr.16: 71475180-71475180 on Build GRCh38
rs138723905 Chr.16: 71475342-71475342 on Build GRCh38
rs367607908 Chr.16: 71475363-71475363 on Build GRCh38
rs764989543 Chr.16: 71475285-71475285 on Build GRCh38
rs780049307 Chr.16: 71475243-71475243 on Build GRCh38
rs1033999016 Chr.16: 71475254-71475254 on Build GRCh38
rs191331007 Chr.16: 71475211-71475211 on Build GRCh38
rs748591073 Chr.16: 71475253-71475253 on Build GRCh38
rs765978266 Chr.16: 71475185-71475185 on Build GRCh38
rs752262390 Chr.16: 71475287-71475287 on Build GRCh38
rs144757223 Chr.16: 71475330-71475330 on Build GRCh38
rs563994416 Chr.16: 71475333-71475333 on Build GRCh38
rs751510251 Chr.16: 71475314-71475314 on Build GRCh38
rs768015169 Chr.16: 71475159-71475159 on Build GRCh38
rs75731908 Chr.16: 71475304-71475304 on Build GRCh38
rs118106739 Chr.16: 71475217-71475217 on Build GRCh38
rs141589678 Chr.16: 71475351-71475351 on Build GRCh38
rs780136487 Chr.16: 71475327-71475327 on Build GRCh38
rs574565928 Chr.16: 71475262-71475262 on Build GRCh38
rs754425182 Chr.16: 71475251-71475251 on Build GRCh38
rs761209018 Chr.16: 71475270-71475270 on Build GRCh38
rs143589546 Chr.16: 71475200-71475200 on Build GRCh38
rs759135804 Chr.16: 71475276-71475276 on Build GRCh38
rs763879278 Chr.16: 71475378-71475378 on Build GRCh38

View Full Product Details