Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 33455915-33456176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TARS
Entrez Gene ID
6897
Gene Name
threonyl-tRNA synthetase
Gene Aliases
ThrRS
UniGene
Hs.481860
Gene Chromosome Location
Chr.5: 33440696-33468091 on Build GRCh38
Transcripts
NM_001258437.1 NM_001258438.1 NM_152295.4 NR_047676.1 NR_047677.1 NR_047678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3787054 c.713G>A p.R238Q Chr.5: 33456021-33456021 on Build GRCh38
3393531 c.699C>G p.N233K Chr.5: 33456007-33456007 on Build GRCh38
6307754 c.722A>T p.N241I Chr.5: 33456030-33456030 on Build GRCh38
164791 c.695A>G p.Y232C Chr.5: 33456003-33456003 on Build GRCh38
3854749 c.761G>A p.C254Y Chr.5: 33456151-33456151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567728628 Chr.5: 33456080-33456080 on Build GRCh38
rs948663895 Chr.5: 33456016-33456016 on Build GRCh38
rs778573895 Chr.5: 33456001-33456001 on Build GRCh38
rs774592689 Chr.5: 33456032-33456032 on Build GRCh38
rs547884304 Chr.5: 33456079-33456079 on Build GRCh38
rs767631538 Chr.5: 33456040-33456040 on Build GRCh38
rs191567567 Chr.5: 33456141-33456141 on Build GRCh38
rs3765144 Chr.5: 33456105-33456105 on Build GRCh38
rs750780482 Chr.5: 33455954-33455954 on Build GRCh38
rs972151438 Chr.5: 33456101-33456101 on Build GRCh38
rs376652221 Chr.5: 33456100-33456100 on Build GRCh38
rs766513834 Chr.5: 33456031-33456031 on Build GRCh38
rs764075809 Chr.5: 33456061-33456061 on Build GRCh38
rs757014362 Chr.5: 33456063-33456063 on Build GRCh38
rs919877095 Chr.5: 33456009-33456009 on Build GRCh38
rs373138685 Chr.5: 33456005-33456005 on Build GRCh38
rs556828655 Chr.5: 33456109-33456109 on Build GRCh38
rs748715234 Chr.5: 33456006-33456006 on Build GRCh38
rs770395787 Chr.5: 33456020-33456020 on Build GRCh38
rs140359049 Chr.5: 33456021-33456021 on Build GRCh38
rs779029303 Chr.5: 33455978-33455980 on Build GRCh38
rs767472025 Chr.5: 33455976-33455976 on Build GRCh38
rs772752312 Chr.5: 33455995-33455995 on Build GRCh38
rs779723175 Chr.5: 33456007-33456007 on Build GRCh38
rs746513807 Chr.5: 33456103-33456103 on Build GRCh38
rs1041873972 Chr.5: 33455966-33455966 on Build GRCh38
rs753591883 Chr.5: 33456062-33456062 on Build GRCh38
rs752739838 Chr.5: 33456050-33456050 on Build GRCh38
rs200414011 Chr.5: 33456135-33456135 on Build GRCh38
rs763126194 Chr.5: 33455960-33455960 on Build GRCh38
rs752580537 Chr.5: 33455985-33455985 on Build GRCh38
rs766620600 Chr.5: 33455961-33455961 on Build GRCh38
rs757620234 Chr.5: 33455953-33455954 on Build GRCh38
rs569594372 Chr.5: 33455972-33455972 on Build GRCh38
rs771276199 Chr.5: 33456110-33456110 on Build GRCh38
rs745368313 Chr.5: 33455987-33455987 on Build GRCh38
rs143600796 Chr.5: 33455999-33455999 on Build GRCh38
rs751603841 Chr.5: 33455971-33455971 on Build GRCh38
rs532309813 Chr.5: 33455992-33455992 on Build GRCh38
rs759633547 Chr.5: 33456038-33456038 on Build GRCh38
rs200556431 Chr.5: 33455995-33455995 on Build GRCh38
rs779753191 Chr.5: 33456102-33456102 on Build GRCh38
rs767719358 Chr.5: 33456149-33456149 on Build GRCh38
rs966284673 Chr.5: 33456094-33456094 on Build GRCh38
rs759791105 Chr.5: 33456143-33456143 on Build GRCh38
rs756108779 Chr.5: 33456051-33456051 on Build GRCh38
rs147399020 Chr.5: 33456046-33456046 on Build GRCh38

View Full Product Details