Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57302620-57302859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNKS1BP1
Entrez Gene ID
85456
Gene Name
tankyrase 1 binding protein 1
Gene Aliases
TAB182
UniGene
Hs.530730
Gene Chromosome Location
Chr.11: 57299629-57324952 on Build GRCh38
Transcripts
NM_033396.2 XM_006718725.3 XM_011545325.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1354734 c.4397A>T p.K1466M Chr.11: 57302745-57302745 on Build GRCh38
5554671 c.4469G>T p.G1490V Chr.11: 57302673-57302673 on Build GRCh38
3809567 c.4506C>T p.D1502D Chr.11: 57302636-57302636 on Build GRCh38
5540611 c.4349C>T p.S1450F Chr.11: 57302793-57302793 on Build GRCh38
5360692 c.4364T>C p.L1455P Chr.11: 57302778-57302778 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747833425 Chr.11: 57302832-57302832 on Build GRCh38
rs778977402 Chr.11: 57302715-57302715 on Build GRCh38
rs781056912 Chr.11: 57302831-57302831 on Build GRCh38
rs780201735 Chr.11: 57302721-57302721 on Build GRCh38
rs751123821 Chr.11: 57302673-57302673 on Build GRCh38
rs370032030 Chr.11: 57302777-57302777 on Build GRCh38
rs765961905 Chr.11: 57302784-57302784 on Build GRCh38
rs138666146 Chr.11: 57302651-57302651 on Build GRCh38
rs762710976 Chr.11: 57302681-57302681 on Build GRCh38
rs758522769 Chr.11: 57302811-57302811 on Build GRCh38
rs562916895 Chr.11: 57302805-57302805 on Build GRCh38
rs777965629 Chr.11: 57302703-57302703 on Build GRCh38
rs959540888 Chr.11: 57302675-57302675 on Build GRCh38
rs761629633 Chr.11: 57302660-57302660 on Build GRCh38
rs375474498 Chr.11: 57302720-57302720 on Build GRCh38
rs764577456 Chr.11: 57302799-57302799 on Build GRCh38
rs545474995 Chr.11: 57302814-57302814 on Build GRCh38
rs766087555 Chr.11: 57302690-57302690 on Build GRCh38
rs202008729 Chr.11: 57302806-57302806 on Build GRCh38
rs990039463 Chr.11: 57302742-57302742 on Build GRCh38
rs760424521 Chr.11: 57302639-57302639 on Build GRCh38
rs768751538 Chr.11: 57302815-57302815 on Build GRCh38
rs776288885 Chr.11: 57302758-57302758 on Build GRCh38
rs564064579 Chr.11: 57302834-57302834 on Build GRCh38
rs1033876258 Chr.11: 57302673-57302673 on Build GRCh38
rs574992277 Chr.11: 57302779-57302779 on Build GRCh38
rs749943500 Chr.11: 57302680-57302680 on Build GRCh38
rs762500818 Chr.11: 57302782-57302782 on Build GRCh38
rs763709386 Chr.11: 57302656-57302658 on Build GRCh38
rs368983075 Chr.11: 57302812-57302812 on Build GRCh38
rs369759558 Chr.11: 57302726-57302726 on Build GRCh38
rs756547288 Chr.11: 57302699-57302699 on Build GRCh38
rs761268899 Chr.11: 57302799-57302799 on Build GRCh38
rs553627122 Chr.11: 57302705-57302705 on Build GRCh38
rs772877770 Chr.11: 57302780-57302780 on Build GRCh38
rs761870534 Chr.11: 57302768-57302768 on Build GRCh38
rs764872515 Chr.11: 57302666-57302666 on Build GRCh38
rs745879669 Chr.11: 57302719-57302719 on Build GRCh38
rs757691432 Chr.11: 57302713-57302713 on Build GRCh38
rs759105527 Chr.11: 57302792-57302792 on Build GRCh38
rs768540383 Chr.11: 57302734-57302734 on Build GRCh38
rs941668273 Chr.11: 57302733-57302733 on Build GRCh38
rs189301745 Chr.11: 57302791-57302791 on Build GRCh38
rs796129568 Chr.11: 57302804-57302804 on Build GRCh38
rs750527030 Chr.11: 57302810-57302810 on Build GRCh38
rs34905018 Chr.11: 57302696-57302696 on Build GRCh38
rs768500251 Chr.11: 57302641-57302641 on Build GRCh38

View Full Product Details