Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66335394-66335663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIN1
Entrez Gene ID
9610
Gene Name
Ras and Rab interactor 1
Gene Aliases
-
UniGene
Hs.1030
Gene Chromosome Location
Chr.11: 66332063-66337235 on Build GRCh38
Transcripts
NM_004292.2 XM_011545399.2 XM_011545400.2 XM_011545401.2 XM_017018587.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2040014 c.542G>A p.G181D Chr.11: 66335412-66335412 on Build GRCh38
4817364 c.156C>T p.L52L Chr.11: 66335483-66335483 on Build GRCh38
6837994 c.424G>C p.V142L Chr.11: 66335641-66335641 on Build GRCh38
4866272 c.223C>A p.L75M Chr.11: 66335416-66335416 on Build GRCh38
4871974 c.232+8C>T p.? Chr.11: 66335399-66335399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775172095 Chr.11: 66335536-66335536 on Build GRCh38
rs777482562 Chr.11: 66335510-66335510 on Build GRCh38
rs374376753 Chr.11: 66335577-66335577 on Build GRCh38
rs1019905192 Chr.11: 66335487-66335489 on Build GRCh38
rs1054652082 Chr.11: 66335433-66335433 on Build GRCh38
rs374091687 Chr.11: 66335463-66335463 on Build GRCh38
rs924629176 Chr.11: 66335440-66335440 on Build GRCh38
rs749579285 Chr.11: 66335506-66335506 on Build GRCh38
rs763487866 Chr.11: 66335539-66335539 on Build GRCh38
rs759656363 Chr.11: 66335635-66335635 on Build GRCh38
rs377583727 Chr.11: 66335570-66335570 on Build GRCh38
rs747948985 Chr.11: 66335629-66335629 on Build GRCh38
rs781207056 Chr.11: 66335484-66335484 on Build GRCh38
rs148226372 Chr.11: 66335610-66335610 on Build GRCh38
rs952106264 Chr.11: 66335638-66335638 on Build GRCh38
rs898313543 Chr.11: 66335569-66335569 on Build GRCh38
rs771727503 Chr.11: 66335633-66335633 on Build GRCh38
rs745472423 Chr.11: 66335585-66335585 on Build GRCh38
rs763999667 Chr.11: 66335639-66335639 on Build GRCh38
rs750484575 Chr.11: 66335480-66335480 on Build GRCh38
rs751977847 Chr.11: 66335459-66335459 on Build GRCh38
rs773628014 Chr.11: 66335630-66335630 on Build GRCh38
rs370997457 Chr.11: 66335532-66335532 on Build GRCh38
rs767698323 Chr.11: 66335476-66335476 on Build GRCh38
rs572187335 Chr.11: 66335612-66335612 on Build GRCh38
rs757539636 Chr.11: 66335470-66335470 on Build GRCh38
rs80130696 Chr.11: 66335544-66335544 on Build GRCh38
rs781064872 Chr.11: 66335578-66335578 on Build GRCh38
rs756196034 Chr.11: 66335481-66335481 on Build GRCh38
rs772281524 Chr.11: 66335617-66335617 on Build GRCh38
rs370189786 Chr.11: 66335597-66335597 on Build GRCh38
rs144581213 Chr.11: 66335602-66335602 on Build GRCh38
rs370524552 Chr.11: 66335574-66335574 on Build GRCh38
rs200852937 Chr.11: 66335561-66335561 on Build GRCh38
rs780202584 Chr.11: 66335497-66335497 on Build GRCh38
rs748035553 Chr.11: 66335505-66335505 on Build GRCh38
rs1007821968 Chr.11: 66335623-66335623 on Build GRCh38
rs748872712 Chr.11: 66335598-66335598 on Build GRCh38
rs143847360 Chr.11: 66335453-66335453 on Build GRCh38
rs1025330818 Chr.11: 66335589-66335589 on Build GRCh38
rs775912256 Chr.11: 66335416-66335416 on Build GRCh38
rs766760672 Chr.11: 66335604-66335605 on Build GRCh38
rs3016763 Chr.11: 66335575-66335575 on Build GRCh38
rs768825021 Chr.11: 66335572-66335572 on Build GRCh38
rs769139792 Chr.11: 66335535-66335535 on Build GRCh38
rs370521634 Chr.11: 66335511-66335511 on Build GRCh38
rs536258525 Chr.11: 66335534-66335534 on Build GRCh38
rs753925206 Chr.11: 66335573-66335573 on Build GRCh38
rs1002501448 Chr.11: 66335552-66335552 on Build GRCh38
rs751326645 Chr.11: 66335576-66335576 on Build GRCh38
rs756387843 Chr.11: 66335564-66335564 on Build GRCh38
rs751671423 Chr.11: 66335482-66335484 on Build GRCh38
rs373989241 Chr.11: 66335559-66335559 on Build GRCh38
rs763577797 Chr.11: 66335452-66335452 on Build GRCh38
rs541119197 Chr.11: 66335634-66335634 on Build GRCh38
rs774851421 Chr.11: 66335545-66335545 on Build GRCh38
rs192812581 Chr.11: 66335520-66335520 on Build GRCh38

View Full Product Details