Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 46769201-46769462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC38A4
Entrez Gene ID
55089
Gene Name
solute carrier family 38 member 4
Gene Aliases
ATA3, NAT3, PAAT, SNAT4
UniGene
Hs.446077
Gene Chromosome Location
Chr.12: 46764761-46832422 on Build GRCh38
Transcripts
NM_001143824.1 NM_018018.4 XM_005268997.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3772622 c.1355G>A p.R452Q Chr.12: 46769373-46769373 on Build GRCh38
3398735 c.1304G>A p.R435H Chr.12: 46769424-46769424 on Build GRCh38
1476570 c.1336C>T p.R446* Chr.12: 46769392-46769392 on Build GRCh38
7287019 c.1380T>C p.L460L Chr.12: 46769348-46769348 on Build GRCh38
939593 c.1403T>C p.V468A Chr.12: 46769325-46769325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763698118 Chr.12: 46769388-46769388 on Build GRCh38
rs776677923 Chr.12: 46769281-46769281 on Build GRCh38
rs767007882 Chr.12: 46769414-46769414 on Build GRCh38
rs756731873 Chr.12: 46769303-46769303 on Build GRCh38
rs945322786 Chr.12: 46769244-46769244 on Build GRCh38
rs144116908 Chr.12: 46769373-46769373 on Build GRCh38
rs375186720 Chr.12: 46769311-46769311 on Build GRCh38
rs915162092 Chr.12: 46769223-46769223 on Build GRCh38
rs148672623 Chr.12: 46769395-46769395 on Build GRCh38
rs146303356 Chr.12: 46769286-46769286 on Build GRCh38
rs770124433 Chr.12: 46769241-46769241 on Build GRCh38
rs751033894 Chr.12: 46769299-46769299 on Build GRCh38
rs769644483 Chr.12: 46769381-46769381 on Build GRCh38
rs139504322 Chr.12: 46769294-46769294 on Build GRCh38
rs759464554 Chr.12: 46769285-46769285 on Build GRCh38
rs142179678 Chr.12: 46769398-46769398 on Build GRCh38
rs768781761 Chr.12: 46769252-46769252 on Build GRCh38
rs542649471 Chr.12: 46769332-46769332 on Build GRCh38
rs968439079 Chr.12: 46769350-46769350 on Build GRCh38
rs938537130 Chr.12: 46769374-46769374 on Build GRCh38
rs774232681 Chr.12: 46769258-46769258 on Build GRCh38
rs144172905 Chr.12: 46769327-46769327 on Build GRCh38
rs769534567 Chr.12: 46769250-46769250 on Build GRCh38
rs776778314 Chr.12: 46769364-46769364 on Build GRCh38
rs750931344 Chr.12: 46769424-46769424 on Build GRCh38
rs775098293 Chr.12: 46769383-46769383 on Build GRCh38
rs756517058 Chr.12: 46769425-46769425 on Build GRCh38
rs573728001 Chr.12: 46769297-46769297 on Build GRCh38
rs748277601 Chr.12: 46769355-46769355 on Build GRCh38
rs779080751 Chr.12: 46769342-46769342 on Build GRCh38
rs749463804 Chr.12: 46769251-46769251 on Build GRCh38
rs570572460 Chr.12: 46769360-46769360 on Build GRCh38
rs775214035 Chr.12: 46769293-46769293 on Build GRCh38
rs200607998 Chr.12: 46769384-46769384 on Build GRCh38
rs756534240 Chr.12: 46769316-46769316 on Build GRCh38
rs770950090 Chr.12: 46769271-46769271 on Build GRCh38
rs750914803 Chr.12: 46769314-46769314 on Build GRCh38
rs988848667 Chr.12: 46769262-46769262 on Build GRCh38
rs1017050848 Chr.12: 46769368-46769368 on Build GRCh38
rs780380180 Chr.12: 46769242-46769242 on Build GRCh38
rs1042338160 Chr.12: 46769270-46769270 on Build GRCh38
rs773973895 Chr.12: 46769391-46769391 on Build GRCh38
rs755105921 Chr.12: 46769336-46769336 on Build GRCh38
rs760710432 Chr.12: 46769268-46769268 on Build GRCh38

View Full Product Details