Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 4565904-4566147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC1A1
Entrez Gene ID
6505
Gene Name
solute carrier family 1 member 1
Gene Aliases
DCBXA, EAAC1, EAAT3, SCZD18
UniGene
Hs.444915
Gene Chromosome Location
Chr.9: 4490427-4587469 on Build GRCh38
Transcripts
NM_004170.5 XM_011518007.1 XM_011518008.2 XM_011518009.2 XM_011518010.1 XM_017015042.1 XM_017015043.1 XM_017014882.1

Gene Symbol
SPATA6L
Entrez Gene ID
55064
Gene Name
spermatogenesis associated 6 like
Gene Aliases
C9orf68, bA6J24.2
UniGene
Hs.179615
Gene Chromosome Location
Chr.9: 4540092-4666674 on Build GRCh38
Transcripts
NM_004170.5 XM_011518007.1 XM_011518008.2 XM_011518009.2 XM_011518010.1 XM_017015042.1 XM_017015043.1 XM_017014882.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3657623 c.452C>T p.P151L Chr.9: 4566058-4566058 on Build GRCh38
5463798 c.453T>C p.P151P Chr.9: 4566059-4566059 on Build GRCh38
6013360 c.441-9C>T p.? Chr.9: 4566038-4566038 on Build GRCh38
285036 c.483+1G>A p.? Chr.9: 4566090-4566090 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs899629786 Chr.9: 4565932-4565932 on Build GRCh38
rs769630449 Chr.9: 4566017-4566023 on Build GRCh38
rs766767482 Chr.9: 4566074-4566074 on Build GRCh38
rs1046816399 Chr.9: 4566059-4566059 on Build GRCh38
rs763619233 Chr.9: 4566113-4566113 on Build GRCh38
rs371266520 Chr.9: 4566093-4566093 on Build GRCh38
rs180737077 Chr.9: 4565959-4565959 on Build GRCh38
rs111709092 Chr.9: 4566009-4566009 on Build GRCh38
rs375340254 Chr.9: 4566100-4566100 on Build GRCh38
rs751000055 Chr.9: 4566118-4566118 on Build GRCh38
rs1022289457 Chr.9: 4565967-4565967 on Build GRCh38
rs774710357 Chr.9: 4566042-4566042 on Build GRCh38
rs1009680943 Chr.9: 4565964-4565964 on Build GRCh38
rs926904826 Chr.9: 4566005-4566005 on Build GRCh38
rs762535064 Chr.9: 4566111-4566111 on Build GRCh38
rs569617321 Chr.9: 4565948-4565948 on Build GRCh38
rs757592902 Chr.9: 4566021-4566021 on Build GRCh38
rs367826671 Chr.9: 4566076-4566076 on Build GRCh38
rs190969328 Chr.9: 4565943-4565943 on Build GRCh38
rs746915087 Chr.9: 4566024-4566024 on Build GRCh38
rs745637813 Chr.9: 4566028-4566028 on Build GRCh38
rs572173547 Chr.9: 4565990-4565990 on Build GRCh38
rs571691749 Chr.9: 4566091-4566091 on Build GRCh38
rs201099557 Chr.9: 4566010-4566010 on Build GRCh38
rs998030701 Chr.9: 4565939-4565939 on Build GRCh38
rs535336916 Chr.9: 4565988-4565988 on Build GRCh38
rs773191619 Chr.9: 4566068-4566068 on Build GRCh38
rs761206305 Chr.9: 4566069-4566069 on Build GRCh38
rs759887327 Chr.9: 4566084-4566084 on Build GRCh38
rs73383440 Chr.9: 4566036-4566036 on Build GRCh38
rs757170835 Chr.9: 4566025-4566025 on Build GRCh38
rs772136743 Chr.9: 4566055-4566055 on Build GRCh38
rs748386266 Chr.9: 4566047-4566047 on Build GRCh38
rs1053303280 Chr.9: 4565977-4565977 on Build GRCh38
rs372235217 Chr.9: 4566019-4566019 on Build GRCh38
rs893283178 Chr.9: 4565978-4565978 on Build GRCh38

View Full Product Details