Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 120866418-120866665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR11
Entrez Gene ID
55717
Gene Name
WD repeat domain 11
Gene Aliases
BRWD2, DR11, HH14, SRI1, WDR15
UniGene
Hs.144447
Gene Chromosome Location
Chr.10: 120851175-120909526 on Build GRCh38
Transcripts
NM_018117.11 XM_005269962.2 XM_005269963.2 XM_005269964.2 XM_017016397.1 XM_017016398.1 XM_017016399.1 XM_017016400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1188067 c.1052G>T p.R351M Chr.10: 120866626-120866626 on Build GRCh38
4174676 c.1003C>T p.P335S Chr.10: 120866577-120866577 on Build GRCh38
303562 c.1066G>A p.V356I Chr.10: 120866640-120866640 on Build GRCh38
2056022 c.1079G>C p.S360T Chr.10: 120866653-120866653 on Build GRCh38
4429349 c.1024G>C p.D342H Chr.10: 120866598-120866598 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567532687 Chr.10: 120866467-120866467 on Build GRCh38
rs778156032 Chr.10: 120866550-120866550 on Build GRCh38
rs565787965 Chr.10: 120866553-120866553 on Build GRCh38
rs952728601 Chr.10: 120866457-120866457 on Build GRCh38
rs559546915 Chr.10: 120866450-120866450 on Build GRCh38
rs368706496 Chr.10: 120866554-120866554 on Build GRCh38
rs760543760 Chr.10: 120866621-120866621 on Build GRCh38
rs745876336 Chr.10: 120866566-120866566 on Build GRCh38
rs142204275 Chr.10: 120866606-120866606 on Build GRCh38
rs554794736 Chr.10: 120866644-120866644 on Build GRCh38
rs187738542 Chr.10: 120866534-120866534 on Build GRCh38
rs368968347 Chr.10: 120866533-120866533 on Build GRCh38
rs980091077 Chr.10: 120866484-120866484 on Build GRCh38
rs569612664 Chr.10: 120866531-120866531 on Build GRCh38
rs754533420 Chr.10: 120866538-120866538 on Build GRCh38
rs182668545 Chr.10: 120866527-120866527 on Build GRCh38
rs780214514 Chr.10: 120866541-120866541 on Build GRCh38
rs748146797 Chr.10: 120866596-120866596 on Build GRCh38
rs558543124 Chr.10: 120866537-120866537 on Build GRCh38
rs759690143 Chr.10: 120866519-120866519 on Build GRCh38
rs774417914 Chr.10: 120866591-120866591 on Build GRCh38
rs768426743 Chr.10: 120866524-120866524 on Build GRCh38
rs781497933 Chr.10: 120866556-120866556 on Build GRCh38
rs771963536 Chr.10: 120866605-120866605 on Build GRCh38
rs776354573 Chr.10: 120866639-120866639 on Build GRCh38
rs930622668 Chr.10: 120866562-120866562 on Build GRCh38
rs34304988 Chr.10: 120866640-120866640 on Build GRCh38
rs758744705 Chr.10: 120866544-120866544 on Build GRCh38
rs769921316 Chr.10: 120866585-120866585 on Build GRCh38
rs867153224 Chr.10: 120866458-120866458 on Build GRCh38
rs2289335 Chr.10: 120866526-120866526 on Build GRCh38
rs765892068 Chr.10: 120866634-120866634 on Build GRCh38

View Full Product Details