Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46257717-46257982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKDREJ
Entrez Gene ID
10343
Gene Name
polycystin (PKD) family receptor for egg jelly
Gene Aliases
-
UniGene
Hs.241383
Gene Chromosome Location
Chr.22: 46255663-46263322 on Build GRCh38
Transcripts
NM_006071.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3379382 c.5341C>A p.L1781I Chr.22: 46257982-46257982 on Build GRCh38
5030767 c.5555_5556delGT p.S1852fs*7 Chr.22: 46257767-46257768 on Build GRCh38
4444873 c.5560A>G p.N1854D Chr.22: 46257763-46257763 on Build GRCh38
3063437 c.5428G>A p.E1810K Chr.22: 46257895-46257895 on Build GRCh38
6162528 c.5391G>A p.V1797V Chr.22: 46257932-46257932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143695558 Chr.22: 46257818-46257818 on Build GRCh38
rs775595205 Chr.22: 46257863-46257863 on Build GRCh38
rs754599047 Chr.22: 46257790-46257790 on Build GRCh38
rs750430335 Chr.22: 46257777-46257777 on Build GRCh38
rs978735396 Chr.22: 46257837-46257837 on Build GRCh38
rs989646453 Chr.22: 46257957-46257957 on Build GRCh38
rs200001840 Chr.22: 46257764-46257764 on Build GRCh38
rs770989149 Chr.22: 46257822-46257822 on Build GRCh38
rs757696701 Chr.22: 46257929-46257929 on Build GRCh38
rs201984857 Chr.22: 46257772-46257772 on Build GRCh38
rs767649250 Chr.22: 46257767-46257767 on Build GRCh38
rs764067686 Chr.22: 46257895-46257895 on Build GRCh38
rs745909178 Chr.22: 46257951-46257951 on Build GRCh38
rs771299501 Chr.22: 46257916-46257916 on Build GRCh38
rs770106827 Chr.22: 46257810-46257810 on Build GRCh38
rs751045720 Chr.22: 46257889-46257889 on Build GRCh38
rs748497201 Chr.22: 46257807-46257807 on Build GRCh38
rs780711089 Chr.22: 46257805-46257805 on Build GRCh38
rs765159802 Chr.22: 46257871-46257871 on Build GRCh38
rs765171239 Chr.22: 46257775-46257775 on Build GRCh38
rs200961868 Chr.22: 46257912-46257912 on Build GRCh38
rs550522880 Chr.22: 46257896-46257896 on Build GRCh38
rs750380958 Chr.22: 46257878-46257878 on Build GRCh38
rs200020318 Chr.22: 46257784-46257784 on Build GRCh38
rs953517367 Chr.22: 46257792-46257792 on Build GRCh38
rs761643254 Chr.22: 46257866-46257866 on Build GRCh38
rs747333656 Chr.22: 46257907-46257907 on Build GRCh38
rs757946625 Chr.22: 46257781-46257781 on Build GRCh38
rs774555407 Chr.22: 46257749-46257749 on Build GRCh38
rs772294297 Chr.22: 46257842-46257842 on Build GRCh38
rs766430224 Chr.22: 46257888-46257888 on Build GRCh38
rs778223179 Chr.22: 46257817-46257817 on Build GRCh38
rs757899901 Chr.22: 46257894-46257894 on Build GRCh38
rs376982011 Chr.22: 46257950-46257950 on Build GRCh38
rs774361645 Chr.22: 46257833-46257833 on Build GRCh38
rs74610409 Chr.22: 46257884-46257884 on Build GRCh38
rs756498080 Chr.22: 46257914-46257914 on Build GRCh38
rs781418001 Chr.22: 46257949-46257950 on Build GRCh38
rs146830478 Chr.22: 46257923-46257923 on Build GRCh38
rs201464325 Chr.22: 46257788-46257788 on Build GRCh38
rs1052398119 Chr.22: 46257905-46257906 on Build GRCh38
rs753605120 Chr.22: 46257768-46257768 on Build GRCh38
rs749671159 Chr.22: 46257927-46257927 on Build GRCh38

View Full Product Details