Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55819042-55819311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEPT14
Entrez Gene ID
346288
Gene Name
septin 14
Gene Aliases
-
UniGene
Hs.453629
Gene Chromosome Location
Chr.7: 55793544-55862789 on Build GRCh38
Transcripts
NM_207366.2 XM_011515373.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7353349 c.170G>C p.W57S Chr.7: 55819141-55819141 on Build GRCh38
4557905 c.113G>A p.G38E Chr.7: 55819198-55819198 on Build GRCh38
5872906 c.757G>A p.E253K Chr.7: 55819187-55819187 on Build GRCh38
6279009 c.752C>T p.T251I Chr.7: 55819192-55819192 on Build GRCh38
7279024 c.725T>G p.L242R Chr.7: 55819219-55819219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113119978 Chr.7: 55819219-55819219 on Build GRCh38
rs746501862 Chr.7: 55819175-55819175 on Build GRCh38
rs535911994 Chr.7: 55819142-55819142 on Build GRCh38
rs779499046 Chr.7: 55819168-55819168 on Build GRCh38
rs773500052 Chr.7: 55819091-55819091 on Build GRCh38
rs534362024 Chr.7: 55819236-55819236 on Build GRCh38
rs778117180 Chr.7: 55819154-55819154 on Build GRCh38
rs73701152 Chr.7: 55819101-55819101 on Build GRCh38
rs761647714 Chr.7: 55819081-55819081 on Build GRCh38
rs868485772 Chr.7: 55819131-55819131 on Build GRCh38
rs555242551 Chr.7: 55819191-55819191 on Build GRCh38
rs771242346 Chr.7: 55819266-55819266 on Build GRCh38
rs367558091 Chr.7: 55819249-55819253 on Build GRCh38
rs185132156 Chr.7: 55819080-55819080 on Build GRCh38
rs753493254 Chr.7: 55819156-55819156 on Build GRCh38
rs768103604 Chr.7: 55819202-55819202 on Build GRCh38
rs755048652 Chr.7: 55819134-55819134 on Build GRCh38
rs372370181 Chr.7: 55819221-55819221 on Build GRCh38
rs568857740 Chr.7: 55819112-55819112 on Build GRCh38
rs375176436 Chr.7: 55819110-55819110 on Build GRCh38
rs762019494 Chr.7: 55819177-55819177 on Build GRCh38
rs376964833 Chr.7: 55819225-55819225 on Build GRCh38
rs751720698 Chr.7: 55819130-55819130 on Build GRCh38
rs903620549 Chr.7: 55819257-55819257 on Build GRCh38
rs773160433 Chr.7: 55819227-55819227 on Build GRCh38
rs200028817 Chr.7: 55819243-55819247 on Build GRCh38
rs368110863 Chr.7: 55819148-55819148 on Build GRCh38
rs758070422 Chr.7: 55819163-55819163 on Build GRCh38
rs768669465 Chr.7: 55819079-55819079 on Build GRCh38
rs756494333 Chr.7: 55819153-55819153 on Build GRCh38
rs765001371 Chr.7: 55819086-55819086 on Build GRCh38
rs112345371 Chr.7: 55819117-55819117 on Build GRCh38
rs781178255 Chr.7: 55819205-55819205 on Build GRCh38

View Full Product Details