Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132635825-132636092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLE
Entrez Gene ID
5426
Gene Name
DNA polymerase epsilon, catalytic subunit
Gene Aliases
CRCS12, FILS, POLE1
UniGene
Hs.524871
Gene Chromosome Location
Chr.12: 132623762-132687524 on Build GRCh38
Transcripts
NM_006231.3 XM_011534795.2 XM_011534797.2 XM_011534798.1 XM_011534802.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6424307 c.5807G>T p.G1936V Chr.12: 132635896-132635896 on Build GRCh38
5361552 c.5795G>A p.R1932H Chr.12: 132635908-132635908 on Build GRCh38
6136335 c.5797A>G p.I1933V Chr.12: 132635906-132635906 on Build GRCh38
5933119 c.5794C>T p.R1932C Chr.12: 132635909-132635909 on Build GRCh38
6967489 c.5734G>A p.E1912K Chr.12: 132635969-132635969 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760495946 Chr.12: 132635958-132635958 on Build GRCh38
rs764439900 Chr.12: 132636000-132636000 on Build GRCh38
rs375198950 Chr.12: 132635934-132635934 on Build GRCh38
rs886313621 Chr.12: 132636008-132636008 on Build GRCh38
rs765804223 Chr.12: 132635880-132635880 on Build GRCh38
rs771980261 Chr.12: 132635942-132635942 on Build GRCh38
rs781099396 Chr.12: 132635927-132635927 on Build GRCh38
rs879254168 Chr.12: 132635909-132635909 on Build GRCh38
rs541999794 Chr.12: 132635961-132635961 on Build GRCh38
rs758916776 Chr.12: 132636063-132636063 on Build GRCh38
rs761722325 Chr.12: 132635867-132635867 on Build GRCh38
rs1005194290 Chr.12: 132635974-132635974 on Build GRCh38
rs749515529 Chr.12: 132636039-132636039 on Build GRCh38
rs767496794 Chr.12: 132635875-132635875 on Build GRCh38
rs375779857 Chr.12: 132635847-132635847 on Build GRCh38
rs944517039 Chr.12: 132636012-132636012 on Build GRCh38
rs746839543 Chr.12: 132636047-132636047 on Build GRCh38
rs369755963 Chr.12: 132635967-132635967 on Build GRCh38
rs762771232 Chr.12: 132635848-132635848 on Build GRCh38
rs764895623 Chr.12: 132635895-132635895 on Build GRCh38
rs770094685 Chr.12: 132635865-132635865 on Build GRCh38
rs549163600 Chr.12: 132635973-132635973 on Build GRCh38
rs201455049 Chr.12: 132635977-132635977 on Build GRCh38
rs778190944 Chr.12: 132635908-132635908 on Build GRCh38
rs769283982 Chr.12: 132636064-132636064 on Build GRCh38
rs769038179 Chr.12: 132636045-132636045 on Build GRCh38
rs547834555 Chr.12: 132636036-132636036 on Build GRCh38
rs369686061 Chr.12: 132635868-132635868 on Build GRCh38
rs751997713 Chr.12: 132636006-132636006 on Build GRCh38
rs770730049 Chr.12: 132636061-132636061 on Build GRCh38
rs562946055 Chr.12: 132635887-132635887 on Build GRCh38
rs373607180 Chr.12: 132635861-132635861 on Build GRCh38
rs150195182 Chr.12: 132635879-132635879 on Build GRCh38
rs756275127 Chr.12: 132636023-132636023 on Build GRCh38
rs753430384 Chr.12: 132635886-132635886 on Build GRCh38
rs757847276 Chr.12: 132635926-132635926 on Build GRCh38
rs5744990 Chr.12: 132635996-132635996 on Build GRCh38
rs113976595 Chr.12: 132635940-132635940 on Build GRCh38
rs115381003 Chr.12: 132635876-132635876 on Build GRCh38
rs770926261 Chr.12: 132635849-132635849 on Build GRCh38
rs775859562 Chr.12: 132635937-132635937 on Build GRCh38
rs371862779 Chr.12: 132635933-132635933 on Build GRCh38
rs745896415 Chr.12: 132636018-132636018 on Build GRCh38
rs377717161 Chr.12: 132635852-132635852 on Build GRCh38
rs758762868 Chr.12: 132635906-132635906 on Build GRCh38
rs781734042 Chr.12: 132636011-132636011 on Build GRCh38
rs5744991 Chr.12: 132635899-132635899 on Build GRCh38
rs930419326 Chr.12: 132635979-132635979 on Build GRCh38
rs752363769 Chr.12: 132635982-132635982 on Build GRCh38
rs773209557 Chr.12: 132635954-132635954 on Build GRCh38
rs202120601 Chr.12: 132635866-132635866 on Build GRCh38
rs780296577 Chr.12: 132635974-132635974 on Build GRCh38
rs371299730 Chr.12: 132635855-132635855 on Build GRCh38
rs5744992 Chr.12: 132635846-132635846 on Build GRCh38
rs879254232 Chr.12: 132635911-132635911 on Build GRCh38
rs757685059 Chr.12: 132636007-132636007 on Build GRCh38
rs368174082 Chr.12: 132635892-132635892 on Build GRCh38
rs753011472 Chr.12: 132635968-132635968 on Build GRCh38
rs372805732 Chr.12: 132636046-132636046 on Build GRCh38
rs1030387176 Chr.12: 132635938-132635938 on Build GRCh38
rs747456028 Chr.12: 132635915-132635915 on Build GRCh38

View Full Product Details