Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52321629-52321872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF480
Entrez Gene ID
147657
Gene Name
zinc finger protein 480
Gene Aliases
-
UniGene
Hs.729317
Gene Chromosome Location
Chr.19: 52297169-52325927 on Build GRCh38
Transcripts
NM_001297624.1 NM_001297625.1 NM_144684.3 XM_011526465.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5453228 c.612A>T p.E204D Chr.19: 52321862-52321862 on Build GRCh38
4154330 c.451T>A p.C151S Chr.19: 52321701-52321701 on Build GRCh38
5464693 c.383T>G p.L128R Chr.19: 52321633-52321633 on Build GRCh38
5464694 c.326T>G p.L109R Chr.19: 52321633-52321633 on Build GRCh38
440161 c.430G>C p.D144H Chr.19: 52321680-52321680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151111132 Chr.19: 52321738-52321738 on Build GRCh38
rs556955800 Chr.19: 52321787-52321787 on Build GRCh38
rs767347980 Chr.19: 52321706-52321706 on Build GRCh38
rs760286091 Chr.19: 52321720-52321720 on Build GRCh38
rs755330296 Chr.19: 52321736-52321736 on Build GRCh38
rs13343641 Chr.19: 52321779-52321779 on Build GRCh38
rs778993253 Chr.19: 52321772-52321772 on Build GRCh38
rs752814232 Chr.19: 52321753-52321753 on Build GRCh38
rs767734941 Chr.19: 52321841-52321841 on Build GRCh38
rs766936668 Chr.19: 52321820-52321820 on Build GRCh38
rs370996313 Chr.19: 52321810-52321810 on Build GRCh38
rs771052101 Chr.19: 52321771-52321771 on Build GRCh38
rs375407066 Chr.19: 52321839-52321839 on Build GRCh38
rs932158669 Chr.19: 52321824-52321824 on Build GRCh38
rs776526189 Chr.19: 52321678-52321678 on Build GRCh38
rs976218899 Chr.19: 52321798-52321798 on Build GRCh38
rs746343562 Chr.19: 52321767-52321767 on Build GRCh38
rs973954228 Chr.19: 52321708-52321708 on Build GRCh38
rs770855733 Chr.19: 52321690-52321690 on Build GRCh38
rs773223002 Chr.19: 52321680-52321680 on Build GRCh38
rs778081349 Chr.19: 52321765-52321765 on Build GRCh38
rs201003978 Chr.19: 52321814-52321814 on Build GRCh38
rs184656017 Chr.19: 52321764-52321764 on Build GRCh38
rs756249128 Chr.19: 52321759-52321759 on Build GRCh38
rs758832647 Chr.19: 52321731-52321731 on Build GRCh38
rs749263973 Chr.19: 52321766-52321766 on Build GRCh38
rs139661910 Chr.19: 52321687-52321687 on Build GRCh38
rs751977031 Chr.19: 52321734-52321734 on Build GRCh38
rs777648657 Chr.19: 52321762-52321762 on Build GRCh38
rs922065067 Chr.19: 52321823-52321823 on Build GRCh38
rs142942426 Chr.19: 52321702-52321702 on Build GRCh38
rs761297544 Chr.19: 52321816-52321816 on Build GRCh38
rs367848964 Chr.19: 52321782-52321782 on Build GRCh38
rs201507498 Chr.19: 52321724-52321724 on Build GRCh38
rs750966916 Chr.19: 52321730-52321730 on Build GRCh38
rs775226352 Chr.19: 52321789-52321789 on Build GRCh38
rs1026917825 Chr.19: 52321710-52321710 on Build GRCh38
rs149769756 Chr.19: 52321826-52321826 on Build GRCh38
rs774977711 Chr.19: 52321718-52321718 on Build GRCh38
rs769729814 Chr.19: 52321658-52321658 on Build GRCh38
rs758086042 Chr.19: 52321689-52321690 on Build GRCh38
rs774232026 Chr.19: 52321691-52321691 on Build GRCh38
rs766740368 Chr.19: 52321732-52321732 on Build GRCh38
rs777407473 Chr.19: 52321661-52321661 on Build GRCh38
rs971575957 Chr.19: 52321673-52321673 on Build GRCh38

View Full Product Details