Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 3452089-3452316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGIF1
Entrez Gene ID
7050
Gene Name
TGFB induced factor homeobox 1
Gene Aliases
HPE4, TGIF
UniGene
Hs.373550
Gene Chromosome Location
Chr.18: 3411927-3458412 on Build GRCh38
Transcripts
NM_001278682.1 NM_001278684.1 NM_001278686.1 NM_003244.3 NM_170695.3 NM_173207.2 NM_173208.2 NM_173209.2 NM_173210.2 NM_174886.2 XM_011525735.2 XM_017025958.1 XM_017025959.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7085030 c.252A>G p.P84P Chr.18: 3452231-3452231 on Build GRCh38
6458624 c.238_239insC p.P83fs*61 Chr.18: 3452217-3452218 on Build GRCh38
296436 c.246delT p.P83fs*51 Chr.18: 3452225-3452225 on Build GRCh38
4565589 c.225_226GG>AA p.G76R Chr.18: 3452204-3452205 on Build GRCh38
4765021 c.239_240insC p.P83fs*61 Chr.18: 3452218-3452219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750230453 Chr.18: 3452158-3452158 on Build GRCh38
rs748158154 Chr.18: 3452149-3452149 on Build GRCh38
rs767498862 Chr.18: 3452127-3452127 on Build GRCh38
rs557930227 Chr.18: 3452217-3452218 on Build GRCh38
rs753529856 Chr.18: 3452191-3452191 on Build GRCh38
rs200428035 Chr.18: 3452190-3452190 on Build GRCh38
rs780619862 Chr.18: 3452218-3452220 on Build GRCh38
rs11571509 Chr.18: 3452218-3452218 on Build GRCh38
rs781655696 Chr.18: 3452290-3452290 on Build GRCh38
rs760499860 Chr.18: 3452188-3452188 on Build GRCh38
rs747161834 Chr.18: 3452254-3452254 on Build GRCh38
rs373376588 Chr.18: 3452173-3452173 on Build GRCh38
rs1028388866 Chr.18: 3452251-3452251 on Build GRCh38
rs765997279 Chr.18: 3452188-3452188 on Build GRCh38
rs368163691 Chr.18: 3452198-3452198 on Build GRCh38
rs757783979 Chr.18: 3452288-3452288 on Build GRCh38
rs780454778 Chr.18: 3452228-3452228 on Build GRCh38
rs757652544 Chr.18: 3452219-3452219 on Build GRCh38
rs750327103 Chr.18: 3452128-3452128 on Build GRCh38
rs543624092 Chr.18: 3452217-3452217 on Build GRCh38
rs559432063 Chr.18: 3452224-3452224 on Build GRCh38
rs772275305 Chr.18: 3452225-3452228 on Build GRCh38
rs150364699 Chr.18: 3452186-3452186 on Build GRCh38
rs549975622 Chr.18: 3452122-3452122 on Build GRCh38
rs557414388 Chr.18: 3452201-3452201 on Build GRCh38
rs1051725877 Chr.18: 3452139-3452139 on Build GRCh38
rs763117453 Chr.18: 3452276-3452276 on Build GRCh38
rs746165812 Chr.18: 3452220-3452220 on Build GRCh38
rs960289467 Chr.18: 3452243-3452243 on Build GRCh38
rs763724267 Chr.18: 3452123-3452123 on Build GRCh38
rs761484867 Chr.18: 3452174-3452174 on Build GRCh38
rs776816609 Chr.18: 3452170-3452170 on Build GRCh38
rs776544188 Chr.18: 3452261-3452261 on Build GRCh38
rs868702917 Chr.18: 3452142-3452142 on Build GRCh38
rs770717714 Chr.18: 3452248-3452248 on Build GRCh38
rs747784486 Chr.18: 3452226-3452226 on Build GRCh38
rs778402135 Chr.18: 3452162-3452162 on Build GRCh38
rs557543525 Chr.18: 3452225-3452225 on Build GRCh38
rs17852936 Chr.18: 3452266-3452266 on Build GRCh38
rs914760425 Chr.18: 3452156-3452157 on Build GRCh38
rs866113270 Chr.18: 3452143-3452143 on Build GRCh38
rs796625425 Chr.18: 3452224-3452225 on Build GRCh38
rs1057417541 Chr.18: 3452192-3452192 on Build GRCh38
rs11571510 Chr.18: 3452225-3452225 on Build GRCh38
rs138752728 Chr.18: 3452206-3452206 on Build GRCh38
rs758522634 Chr.18: 3452209-3452209 on Build GRCh38
rs758465805 Chr.18: 3452157-3452157 on Build GRCh38
rs764252611 Chr.18: 3452287-3452287 on Build GRCh38
rs931796360 Chr.18: 3452296-3452296 on Build GRCh38
rs1035910449 Chr.18: 3452238-3452238 on Build GRCh38
rs760686702 Chr.18: 3452232-3452232 on Build GRCh38
rs768870784 Chr.18: 3452255-3452256 on Build GRCh38
rs761167003 Chr.18: 3452114-3452114 on Build GRCh38
rs543310164 Chr.18: 3452271-3452271 on Build GRCh38
rs8098291 Chr.18: 3452264-3452264 on Build GRCh38
rs202189171 Chr.18: 3452224-3452225 on Build GRCh38
rs868610906 Chr.18: 3452110-3452110 on Build GRCh38
rs1002110854 Chr.18: 3452228-3452228 on Build GRCh38
rs765001208 Chr.18: 3452137-3452137 on Build GRCh38
rs761753994 Chr.18: 3452125-3452128 on Build GRCh38
rs112625272 Chr.18: 3452185-3452185 on Build GRCh38
rs770281321 Chr.18: 3452222-3452222 on Build GRCh38
rs555016322 Chr.18: 3452164-3452164 on Build GRCh38
rs147815261 Chr.18: 3452134-3452134 on Build GRCh38
rs114378990 Chr.18: 3452137-3452137 on Build GRCh38
rs121909070 Chr.18: 3452156-3452156 on Build GRCh38
rs762555858 Chr.18: 3452187-3452188 on Build GRCh38
rs773451816 Chr.18: 3452112-3452112 on Build GRCh38
rs752956314 Chr.18: 3452272-3452272 on Build GRCh38
rs773947728 Chr.18: 3452124-3452124 on Build GRCh38
rs199536188 Chr.18: 3452293-3452293 on Build GRCh38
rs569823526 Chr.18: 3452125-3452125 on Build GRCh38
rs755998929 Chr.18: 3452129-3452129 on Build GRCh38
rs757775729 Chr.18: 3452223-3452225 on Build GRCh38
rs879008093 Chr.18: 3452153-3452153 on Build GRCh38
rs747449695 Chr.18: 3452163-3452163 on Build GRCh38
rs372356142 Chr.18: 3452295-3452295 on Build GRCh38
rs754129491 Chr.18: 3452131-3452131 on Build GRCh38
rs766065938 Chr.18: 3452130-3452130 on Build GRCh38
rs375741920 Chr.18: 3452118-3452118 on Build GRCh38
rs777683230 Chr.18: 3452210-3452210 on Build GRCh38

View Full Product Details