Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128276276-128276513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SWI5
Entrez Gene ID
375757
Gene Name
SWI5 homologous recombination repair protein
Gene Aliases
C9orf119, SAE3
UniGene
Hs.259594
Gene Chromosome Location
Chr.9: 128275379-128288989 on Build GRCh38
Transcripts
NM_001040011.1 NM_001318089.1 NM_001318092.1 XM_011518652.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
216126 c.178C>G p.R60G Chr.9: 128276323-128276323 on Build GRCh38
3904399 c.184C>T p.P62S Chr.9: 128276329-128276329 on Build GRCh38
1489695 c.158G>A p.G53D Chr.9: 128276303-128276303 on Build GRCh38
1733115 c.215T>C p.L72S Chr.9: 128276360-128276360 on Build GRCh38
6299543 c.205C>A p.L69I Chr.9: 128276350-128276350 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888159675 Chr.9: 128276392-128276392 on Build GRCh38
rs766938158 Chr.9: 128276313-128276313 on Build GRCh38
rs749888729 Chr.9: 128276314-128276314 on Build GRCh38
rs374353743 Chr.9: 128276312-128276312 on Build GRCh38
rs540264998 Chr.9: 128276338-128276338 on Build GRCh38
rs1041343182 Chr.9: 128276360-128276360 on Build GRCh38
rs562691586 Chr.9: 128276443-128276443 on Build GRCh38
rs759122854 Chr.9: 128276419-128276419 on Build GRCh38
rs747867413 Chr.9: 128276378-128276386 on Build GRCh38
rs774205594 Chr.9: 128276351-128276358 on Build GRCh38
rs780045741 Chr.9: 128276489-128276489 on Build GRCh38
rs901037600 Chr.9: 128276363-128276363 on Build GRCh38
rs747263258 Chr.9: 128276327-128276327 on Build GRCh38
rs199501836 Chr.9: 128276358-128276358 on Build GRCh38
rs978957695 Chr.9: 128276306-128276306 on Build GRCh38
rs569075444 Chr.9: 128276297-128276297 on Build GRCh38
rs115231472 Chr.9: 128276296-128276296 on Build GRCh38
rs201725177 Chr.9: 128276332-128276332 on Build GRCh38
rs771102225 Chr.9: 128276337-128276337 on Build GRCh38
rs760301673 Chr.9: 128276315-128276315 on Build GRCh38
rs756652422 Chr.9: 128276384-128276384 on Build GRCh38
rs771174251 Chr.9: 128276304-128276304 on Build GRCh38
rs752635793 Chr.9: 128276325-128276325 on Build GRCh38
rs1053681072 Chr.9: 128276353-128276353 on Build GRCh38
rs765031453 Chr.9: 128276437-128276437 on Build GRCh38
rs770310130 Chr.9: 128276345-128276345 on Build GRCh38
rs376149218 Chr.9: 128276344-128276344 on Build GRCh38
rs553787072 Chr.9: 128276347-128276347 on Build GRCh38
rs899649511 Chr.9: 128276417-128276417 on Build GRCh38
rs376809666 Chr.9: 128276453-128276453 on Build GRCh38
rs79146692 Chr.9: 128276488-128276488 on Build GRCh38
rs759953432 Chr.9: 128276310-128276310 on Build GRCh38
rs983746553 Chr.9: 128276301-128276301 on Build GRCh38
rs748029621 Chr.9: 128276295-128276295 on Build GRCh38
rs766049606 Chr.9: 128276316-128276316 on Build GRCh38
rs1033990829 Chr.9: 128276465-128276465 on Build GRCh38
rs747111566 Chr.9: 128276336-128276336 on Build GRCh38
rs73672486 Chr.9: 128276364-128276364 on Build GRCh38
rs200812021 Chr.9: 128276323-128276323 on Build GRCh38
rs749770444 Chr.9: 128276354-128276354 on Build GRCh38
rs971289652 Chr.9: 128276326-128276326 on Build GRCh38
rs144191558 Chr.9: 128276311-128276311 on Build GRCh38
rs776296578 Chr.9: 128276404-128276404 on Build GRCh38

View Full Product Details