Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 55234723-55234978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB27A
Entrez Gene ID
5873
Gene Name
RAB27A, member RAS oncogene family
Gene Aliases
GS2, HsT18676, RAB27, RAM
UniGene
Hs.626665
Gene Chromosome Location
Chr.15: 55202966-55291188 on Build GRCh38
Transcripts
NM_004580.4 NM_183234.2 NM_183235.2 NM_183236.2 XM_005254576.4 XM_011521852.1 XM_011521854.1 XM_011521855.2 XM_011521856.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6761328 c.148_149insA p.R50fs*34 Chr.15: 55234786-55234787 on Build GRCh38
5971466 c.53C>G p.S18C Chr.15: 55234882-55234882 on Build GRCh38
1608489 c.1A>G p.M1V Chr.15: 55234934-55234934 on Build GRCh38
4922023 c.123A>T p.T41T Chr.15: 55234812-55234812 on Build GRCh38
3731160 c.32A>G p.K11R Chr.15: 55234903-55234903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749531175 Chr.15: 55234915-55234915 on Build GRCh38
rs756730276 Chr.15: 55234838-55234838 on Build GRCh38
rs539575657 Chr.15: 55234924-55234924 on Build GRCh38
rs145253993 Chr.15: 55234918-55234918 on Build GRCh38
rs998047728 Chr.15: 55234842-55234842 on Build GRCh38
rs770011320 Chr.15: 55234759-55234759 on Build GRCh38
rs927639143 Chr.15: 55234758-55234758 on Build GRCh38
rs369478092 Chr.15: 55234877-55234877 on Build GRCh38
rs150782434 Chr.15: 55234843-55234843 on Build GRCh38
rs905779372 Chr.15: 55234898-55234898 on Build GRCh38
rs773341484 Chr.15: 55234760-55234760 on Build GRCh38
rs746770347 Chr.15: 55234864-55234864 on Build GRCh38
rs1001351367 Chr.15: 55234900-55234900 on Build GRCh38
rs375882552 Chr.15: 55234768-55234768 on Build GRCh38
rs369152309 Chr.15: 55234818-55234818 on Build GRCh38
rs763762181 Chr.15: 55234805-55234805 on Build GRCh38
rs777995262 Chr.15: 55234886-55234886 on Build GRCh38
rs1024213806 Chr.15: 55234894-55234894 on Build GRCh38
rs746673176 Chr.15: 55234944-55234944 on Build GRCh38
rs758381859 Chr.15: 55234848-55234848 on Build GRCh38
rs142212580 Chr.15: 55234788-55234788 on Build GRCh38
rs778448317 Chr.15: 55234841-55234841 on Build GRCh38
rs753437628 Chr.15: 55234830-55234830 on Build GRCh38
rs141281020 Chr.15: 55234933-55234933 on Build GRCh38
rs760188894 Chr.15: 55234776-55234776 on Build GRCh38
rs952920589 Chr.15: 55234920-55234920 on Build GRCh38
rs376130474 Chr.15: 55234946-55234946 on Build GRCh38
rs770601673 Chr.15: 55234786-55234786 on Build GRCh38
rs553316776 Chr.15: 55234937-55234937 on Build GRCh38
rs139145499 Chr.15: 55234850-55234850 on Build GRCh38
rs200241598 Chr.15: 55234851-55234851 on Build GRCh38
rs1131388 Chr.15: 55234792-55234792 on Build GRCh38
rs768784784 Chr.15: 55234879-55234879 on Build GRCh38
rs770930489 Chr.15: 55234770-55234770 on Build GRCh38
rs767963780 Chr.15: 55234787-55234787 on Build GRCh38
rs774986160 Chr.15: 55234772-55234772 on Build GRCh38

View Full Product Details