Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133412262-133412531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REXO4
Entrez Gene ID
57109
Gene Name
REX4 homolog, 3'-5' exonuclease
Gene Aliases
REX4, XPMC2, XPMC2H
UniGene
Hs.438409
Gene Chromosome Location
Chr.9: 133406058-133418172 on Build GRCh38
Transcripts
NM_001279349.1 NM_001279350.1 NM_001279351.1 NM_020385.3 NR_103995.1 NR_103996.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4371801 c.875C>T p.A292V Chr.9: 133412334-133412334 on Build GRCh38
6114918 c.810G>T p.Q270H Chr.9: 133412399-133412399 on Build GRCh38
7208315 c.789C>A p.A263A Chr.9: 133412420-133412420 on Build GRCh38
3905291 c.910+2T>C p.? Chr.9: 133412297-133412297 on Build GRCh38
4721128 c.756C>T p.G252G Chr.9: 133412453-133412453 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782503520 Chr.9: 133412495-133412495 on Build GRCh38
rs782776107 Chr.9: 133412410-133412410 on Build GRCh38
rs782745257 Chr.9: 133412454-133412454 on Build GRCh38
rs782759821 Chr.9: 133412313-133412313 on Build GRCh38
rs782780392 Chr.9: 133412387-133412387 on Build GRCh38
rs781905704 Chr.9: 133412488-133412488 on Build GRCh38
rs782759442 Chr.9: 133412393-133412393 on Build GRCh38
rs782390204 Chr.9: 133412353-133412353 on Build GRCh38
rs782582308 Chr.9: 133412418-133412418 on Build GRCh38
rs139021248 Chr.9: 133412319-133412319 on Build GRCh38
rs376245202 Chr.9: 133412428-133412428 on Build GRCh38
rs915484404 Chr.9: 133412379-133412379 on Build GRCh38
rs372977426 Chr.9: 133412440-133412440 on Build GRCh38
rs376343918 Chr.9: 133412423-133412423 on Build GRCh38
rs372666425 Chr.9: 133412372-133412372 on Build GRCh38
rs782000675 Chr.9: 133412453-133412453 on Build GRCh38
rs782036354 Chr.9: 133412472-133412477 on Build GRCh38
rs781961926 Chr.9: 133412438-133412438 on Build GRCh38
rs782589633 Chr.9: 133412354-133412354 on Build GRCh38
rs782154359 Chr.9: 133412448-133412448 on Build GRCh38
rs782175150 Chr.9: 133412309-133412309 on Build GRCh38
rs782192693 Chr.9: 133412422-133412422 on Build GRCh38
rs2285486 Chr.9: 133412360-133412360 on Build GRCh38
rs782750666 Chr.9: 133412381-133412381 on Build GRCh38
rs782398032 Chr.9: 133412430-133412430 on Build GRCh38
rs782715873 Chr.9: 133412289-133412289 on Build GRCh38
rs2285487 Chr.9: 133412362-133412362 on Build GRCh38
rs782657874 Chr.9: 133412497-133412497 on Build GRCh38
rs762447422 Chr.9: 133412320-133412320 on Build GRCh38
rs782229177 Chr.9: 133412355-133412355 on Build GRCh38
rs782681565 Chr.9: 133412348-133412348 on Build GRCh38
rs782528793 Chr.9: 133412334-133412334 on Build GRCh38
rs370225194 Chr.9: 133412452-133412452 on Build GRCh38
rs782574183 Chr.9: 133412326-133412326 on Build GRCh38
rs781794727 Chr.9: 133412456-133412456 on Build GRCh38
rs997943579 Chr.9: 133412434-133412434 on Build GRCh38
rs374473489 Chr.9: 133412485-133412485 on Build GRCh38
rs781905350 Chr.9: 133412407-133412407 on Build GRCh38
rs200413290 Chr.9: 133412386-133412386 on Build GRCh38
rs1017292757 Chr.9: 133412443-133412443 on Build GRCh38
rs143153905 Chr.9: 133412329-133412329 on Build GRCh38
rs782227191 Chr.9: 133412408-133412408 on Build GRCh38
rs201264176 Chr.9: 133412359-133412359 on Build GRCh38
rs587768877 Chr.9: 133412371-133412371 on Build GRCh38
rs182537099 Chr.9: 133412291-133412291 on Build GRCh38
rs199826687 Chr.9: 133412464-133412464 on Build GRCh38
rs782245135 Chr.9: 133412349-133412349 on Build GRCh38
rs376943789 Chr.9: 133412441-133412441 on Build GRCh38

View Full Product Details