Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 24127334-24127579 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THRB
Entrez Gene ID
7068
Gene Name
thyroid hormone receptor beta
Gene Aliases
C-ERBA-2, C-ERBA-BETA, ERBA2, GRTH, NR1A2, PRTH, THR1, THRB1, THRB2
UniGene
Hs.187861
Gene Chromosome Location
Chr.3: 24117153-24495281 on Build GRCh38
Transcripts
NM_000461.4 NM_001128176.2 NM_001128177.1 NM_001252634.1 XM_005265421.4 XM_005265423.4 XM_005265424.3 XM_006713317.3 XM_006713318.3 XM_011534046.2 XM_011534047.2 XM_011534048.2 XM_011534049.2 XM_011534050.2 XM_011534051.2 XM_011534052.2 XM_011534053.2 XM_017007108.1 XM_017007109.1 XM_017007110.1 XM_017007111.1 XM_017007112.1 XM_017007113.1 XM_017007114.1 XM_017007115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
446308 c.1099G>A p.D367N Chr.3: 24127544-24127544 on Build GRCh38
4618325 c.1078C>A p.L360M Chr.3: 24127565-24127565 on Build GRCh38
5465830 c.1085C>A p.S362Y Chr.3: 24127558-24127558 on Build GRCh38
6201384 c.1144+6T>C p.? Chr.3: 24127493-24127493 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1037149521 Chr.3: 24127432-24127432 on Build GRCh38
rs796887899 Chr.3: 24127480-24127481 on Build GRCh38
rs756497285 Chr.3: 24127550-24127550 on Build GRCh38
rs762924308 Chr.3: 24127517-24127517 on Build GRCh38
rs963479612 Chr.3: 24127446-24127446 on Build GRCh38
rs80341144 Chr.3: 24127480-24127480 on Build GRCh38
rs942622694 Chr.3: 24127363-24127363 on Build GRCh38
rs753586940 Chr.3: 24127459-24127459 on Build GRCh38
rs541526566 Chr.3: 24127467-24127467 on Build GRCh38
rs148991421 Chr.3: 24127518-24127518 on Build GRCh38
rs900855197 Chr.3: 24127403-24127403 on Build GRCh38
rs780753772 Chr.3: 24127539-24127539 on Build GRCh38
rs750405806 Chr.3: 24127496-24127496 on Build GRCh38
rs778288634 Chr.3: 24127451-24127451 on Build GRCh38
rs1048347326 Chr.3: 24127374-24127374 on Build GRCh38
rs10611180 Chr.3: 24127371-24127374 on Build GRCh38
rs150960035 Chr.3: 24127536-24127536 on Build GRCh38
rs909924770 Chr.3: 24127404-24127404 on Build GRCh38
rs761738765 Chr.3: 24127481-24127481 on Build GRCh38
rs1005047506 Chr.3: 24127434-24127434 on Build GRCh38
rs73825918 Chr.3: 24127423-24127423 on Build GRCh38
rs997834407 Chr.3: 24127420-24127420 on Build GRCh38
rs749748932 Chr.3: 24127457-24127457 on Build GRCh38
rs752310564 Chr.3: 24127540-24127540 on Build GRCh38
rs13063628 Chr.3: 24127490-24127490 on Build GRCh38
rs932918667 Chr.3: 24127358-24127358 on Build GRCh38
rs189421185 Chr.3: 24127465-24127465 on Build GRCh38
rs765060257 Chr.3: 24127504-24127504 on Build GRCh38
rs770116510 Chr.3: 24127449-24127449 on Build GRCh38
rs751094559 Chr.3: 24127521-24127521 on Build GRCh38
rs1016377913 Chr.3: 24127444-24127444 on Build GRCh38
rs558966424 Chr.3: 24127461-24127461 on Build GRCh38
rs370206640 Chr.3: 24127491-24127491 on Build GRCh38
rs772240747 Chr.3: 24127476-24127476 on Build GRCh38
rs184529094 Chr.3: 24127412-24127412 on Build GRCh38
rs897635648 Chr.3: 24127445-24127445 on Build GRCh38
rs886841289 Chr.3: 24127427-24127427 on Build GRCh38

View Full Product Details