Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48113218-48113485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX11
Entrez Gene ID
29916
Gene Name
sorting nexin 11
Gene Aliases
-
UniGene
Hs.15827
Gene Chromosome Location
Chr.17: 48107533-48123064 on Build GRCh38
Transcripts
NM_013323.2 NM_152244.1 XM_005257260.3 XM_005257261.3 XM_005257262.3 XM_011524697.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2697280 c.189C>T p.F63F Chr.17: 48113360-48113360 on Build GRCh38
3362189 c.169C>T p.R57W Chr.17: 48113340-48113340 on Build GRCh38
6786388 c.182G>A p.R61H Chr.17: 48113353-48113353 on Build GRCh38
5621878 c.230T>G p.V77G Chr.17: 48113401-48113401 on Build GRCh38
5055551 c.175C>T p.R59C Chr.17: 48113346-48113346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753586513 Chr.17: 48113379-48113379 on Build GRCh38
rs200037794 Chr.17: 48113254-48113254 on Build GRCh38
rs201964384 Chr.17: 48113440-48113440 on Build GRCh38
rs987950317 Chr.17: 48113418-48113418 on Build GRCh38
rs759016460 Chr.17: 48113261-48113266 on Build GRCh38
rs779711449 Chr.17: 48113361-48113361 on Build GRCh38
rs183763955 Chr.17: 48113424-48113424 on Build GRCh38
rs369398524 Chr.17: 48113353-48113353 on Build GRCh38
rs763096159 Chr.17: 48113265-48113265 on Build GRCh38
rs757237284 Chr.17: 48113278-48113278 on Build GRCh38
rs781172848 Chr.17: 48113283-48113283 on Build GRCh38
rs775693261 Chr.17: 48113422-48113422 on Build GRCh38
rs755941935 Chr.17: 48113358-48113358 on Build GRCh38
rs761997913 Chr.17: 48113451-48113451 on Build GRCh38
rs752801929 Chr.17: 48113257-48113257 on Build GRCh38
rs967457583 Chr.17: 48113394-48113394 on Build GRCh38
rs925932536 Chr.17: 48113456-48113456 on Build GRCh38
rs761942824 Chr.17: 48113350-48113350 on Build GRCh38
rs188333158 Chr.17: 48113300-48113300 on Build GRCh38
rs746351973 Chr.17: 48113411-48113411 on Build GRCh38
rs767430635 Chr.17: 48113453-48113453 on Build GRCh38
rs776968212 Chr.17: 48113315-48113315 on Build GRCh38
rs767062922 Chr.17: 48113276-48113276 on Build GRCh38
rs751602131 Chr.17: 48113276-48113276 on Build GRCh38
rs960742376 Chr.17: 48113341-48113341 on Build GRCh38
rs1033623192 Chr.17: 48113313-48113313 on Build GRCh38
rs148968159 Chr.17: 48113322-48113322 on Build GRCh38
rs564643854 Chr.17: 48113446-48113446 on Build GRCh38
rs368194150 Chr.17: 48113297-48113297 on Build GRCh38
rs200632105 Chr.17: 48113320-48113320 on Build GRCh38
rs1016575553 Chr.17: 48113272-48113272 on Build GRCh38
rs376166068 Chr.17: 48113294-48113294 on Build GRCh38
rs775844175 Chr.17: 48113340-48113340 on Build GRCh38
rs765569909 Chr.17: 48113450-48113450 on Build GRCh38
rs747691853 Chr.17: 48113395-48113395 on Build GRCh38
rs757858237 Chr.17: 48113406-48113406 on Build GRCh38
rs777181961 Chr.17: 48113376-48113376 on Build GRCh38
rs749560489 Chr.17: 48113427-48113427 on Build GRCh38
rs147076627 Chr.17: 48113352-48113352 on Build GRCh38
rs369203100 Chr.17: 48113266-48113266 on Build GRCh38
rs778437886 Chr.17: 48113388-48113388 on Build GRCh38
rs191229840 Chr.17: 48113309-48113309 on Build GRCh38
rs8080662 Chr.17: 48113457-48113457 on Build GRCh38
rs770250111 Chr.17: 48113421-48113421 on Build GRCh38
rs762291905 Chr.17: 48113389-48113390 on Build GRCh38
rs781722907 Chr.17: 48113409-48113409 on Build GRCh38
rs370790863 Chr.17: 48113347-48113347 on Build GRCh38
rs754605681 Chr.17: 48113383-48113383 on Build GRCh38
rs768224550 Chr.17: 48113303-48113303 on Build GRCh38
rs773810584 Chr.17: 48113252-48113253 on Build GRCh38
rs764260449 Chr.17: 48113269-48113269 on Build GRCh38
rs917303731 Chr.17: 48113390-48113390 on Build GRCh38
rs960817124 Chr.17: 48113314-48113314 on Build GRCh38
rs778382822 Chr.17: 48113306-48113306 on Build GRCh38
rs759226849 Chr.17: 48113343-48113343 on Build GRCh38
rs764315505 Chr.17: 48113346-48113346 on Build GRCh38

View Full Product Details