Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74232003-74232268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFC2
Entrez Gene ID
5982
Gene Name
replication factor C subunit 2
Gene Aliases
RFC40
UniGene
Hs.647062
Gene Chromosome Location
Chr.7: 74231502-74254458 on Build GRCh38
Transcripts
NM_001278791.1 NM_001278792.1 NM_001278793.1 NM_002914.4 NM_181471.2 XM_006716080.2 XM_017012492.1 XM_017012493.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3640473 c.984G>T p.A328A Chr.7: 74232187-74232187 on Build GRCh38
7000403 c.955-9G>T p.? Chr.7: 74232225-74232225 on Build GRCh38
3640472 c.1051C>T p.P351S Chr.7: 74232120-74232120 on Build GRCh38
6577188 c.1038G>C p.Q346H Chr.7: 74232133-74232133 on Build GRCh38
7106968 c.1015G>A p.G339S Chr.7: 74232156-74232156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893266623 Chr.7: 74232158-74232158 on Build GRCh38
rs782380757 Chr.7: 74232116-74232116 on Build GRCh38
rs921713706 Chr.7: 74232026-74232026 on Build GRCh38
rs781899980 Chr.7: 74232205-74232205 on Build GRCh38
rs782431526 Chr.7: 74232119-74232119 on Build GRCh38
rs41272921 Chr.7: 74232188-74232188 on Build GRCh38
rs139636296 Chr.7: 74232119-74232119 on Build GRCh38
rs782305885 Chr.7: 74232126-74232126 on Build GRCh38
rs782452573 Chr.7: 74232203-74232203 on Build GRCh38
rs547916230 Chr.7: 74232052-74232052 on Build GRCh38
rs368987624 Chr.7: 74232197-74232197 on Build GRCh38
rs782637865 Chr.7: 74232219-74232219 on Build GRCh38
rs782676909 Chr.7: 74232070-74232070 on Build GRCh38
rs535932724 Chr.7: 74232051-74232051 on Build GRCh38
rs782509382 Chr.7: 74232218-74232218 on Build GRCh38
rs782482287 Chr.7: 74232231-74232231 on Build GRCh38
rs782261352 Chr.7: 74232110-74232110 on Build GRCh38
rs3135712 Chr.7: 74232049-74232049 on Build GRCh38
rs782006697 Chr.7: 74232159-74232159 on Build GRCh38
rs782211144 Chr.7: 74232225-74232225 on Build GRCh38
rs149316712 Chr.7: 74232139-74232139 on Build GRCh38
rs372771318 Chr.7: 74232068-74232068 on Build GRCh38
rs782807091 Chr.7: 74232204-74232204 on Build GRCh38
rs181963235 Chr.7: 74232242-74232242 on Build GRCh38
rs782225616 Chr.7: 74232125-74232125 on Build GRCh38
rs777346846 Chr.7: 74232191-74232191 on Build GRCh38
rs41553218 Chr.7: 74232190-74232190 on Build GRCh38
rs377161147 Chr.7: 74232187-74232187 on Build GRCh38
rs782022750 Chr.7: 74232133-74232133 on Build GRCh38
rs984678837 Chr.7: 74232050-74232050 on Build GRCh38
rs782759945 Chr.7: 74232150-74232150 on Build GRCh38
rs1046103167 Chr.7: 74232092-74232092 on Build GRCh38

View Full Product Details