Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2645454-2645699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC34
Entrez Gene ID
100287898
Gene Name
tetratricopeptide repeat domain 34
Gene Aliases
-
UniGene
Hs.632363
Gene Chromosome Location
Chr.1: 2636988-2789737 on Build GRCh38
Transcripts
NM_001242672.1 XM_016999990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6841170 c.752G>A p.R251H Chr.1: 2645499-2645499 on Build GRCh38
7300030 c.733A>G p.T245A Chr.1: 2645518-2645518 on Build GRCh38
6390161 c.719A>G p.K240R Chr.1: 2645532-2645532 on Build GRCh38
6401117 c.788A>G p.Q263R Chr.1: 2645463-2645463 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs937748180 Chr.1: 2645591-2645591 on Build GRCh38
rs146924199 Chr.1: 2645581-2645581 on Build GRCh38
rs575007693 Chr.1: 2645499-2645499 on Build GRCh38
rs1044929693 Chr.1: 2645510-2645510 on Build GRCh38
rs564924875 Chr.1: 2645585-2645585 on Build GRCh38
rs780714549 Chr.1: 2645595-2645595 on Build GRCh38
rs781725254 Chr.1: 2645584-2645584 on Build GRCh38
rs896281413 Chr.1: 2645615-2645615 on Build GRCh38
rs914191836 Chr.1: 2645551-2645551 on Build GRCh38
rs758505892 Chr.1: 2645558-2645558 on Build GRCh38
rs996734552 Chr.1: 2645560-2645560 on Build GRCh38
rs1046850219 Chr.1: 2645544-2645544 on Build GRCh38
rs537936964 Chr.1: 2645477-2645477 on Build GRCh38
rs530770977 Chr.1: 2645614-2645614 on Build GRCh38
rs554219727 Chr.1: 2645545-2645545 on Build GRCh38
rs768127990 Chr.1: 2645487-2645487 on Build GRCh38
rs898453827 Chr.1: 2645656-2645656 on Build GRCh38
rs989460504 Chr.1: 2645596-2645596 on Build GRCh38
rs765198242 Chr.1: 2645654-2645654 on Build GRCh38
rs1004460484 Chr.1: 2645632-2645632 on Build GRCh38
rs77821602 Chr.1: 2645486-2645486 on Build GRCh38
rs915141641 Chr.1: 2645606-2645606 on Build GRCh38
rs781295222 Chr.1: 2645557-2645557 on Build GRCh38
rs573008055 Chr.1: 2645546-2645546 on Build GRCh38
rs914781874 Chr.1: 2645576-2645576 on Build GRCh38
rs988320704 Chr.1: 2645521-2645521 on Build GRCh38
rs777795717 Chr.1: 2645580-2645580 on Build GRCh38
rs927779674 Chr.1: 2645624-2645624 on Build GRCh38
rs770389540 Chr.1: 2645592-2645592 on Build GRCh38
rs750327662 Chr.1: 2645527-2645527 on Build GRCh38
rs28547181 Chr.1: 2645520-2645520 on Build GRCh38
rs761501123 Chr.1: 2645514-2645514 on Build GRCh38
rs760653494 Chr.1: 2645540-2645540 on Build GRCh38
rs946383713 Chr.1: 2645556-2645556 on Build GRCh38

View Full Product Details