Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95433195-95433456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNE3
Entrez Gene ID
161176
Gene Name
spectrin repeat containing nuclear envelope family member 3
Gene Aliases
C14orf49, NET53, Nesp3
UniGene
Hs.354645
Gene Chromosome Location
Chr.14: 95416083-95516693 on Build GRCh38
Transcripts
NM_152592.3 XM_005267377.3 XM_006720063.3 XM_011536515.2 XM_017021056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5440472 c.2591G>A p.R864H Chr.14: 95433357-95433357 on Build GRCh38
6554668 c.2614T>A p.S872T Chr.14: 95433334-95433334 on Build GRCh38
6554669 c.2614T>A p.S872T Chr.14: 95433334-95433334 on Build GRCh38
6276017 c.2600C>A p.P867Q Chr.14: 95433348-95433348 on Build GRCh38
2253836 c.2641C>T p.Q881* Chr.14: 95433307-95433307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987276582 Chr.14: 95433306-95433306 on Build GRCh38
rs542033506 Chr.14: 95433397-95433397 on Build GRCh38
rs1026008556 Chr.14: 95433260-95433260 on Build GRCh38
rs141951711 Chr.14: 95433358-95433358 on Build GRCh38
rs1019117957 Chr.14: 95433392-95433392 on Build GRCh38
rs529915336 Chr.14: 95433396-95433396 on Build GRCh38
rs569529532 Chr.14: 95433310-95433310 on Build GRCh38
rs748587924 Chr.14: 95433397-95433398 on Build GRCh38
rs773278706 Chr.14: 95433284-95433284 on Build GRCh38
rs748245892 Chr.14: 95433277-95433277 on Build GRCh38
rs111285231 Chr.14: 95433278-95433278 on Build GRCh38
rs150194117 Chr.14: 95433335-95433335 on Build GRCh38
rs769804447 Chr.14: 95433376-95433376 on Build GRCh38
rs765029810 Chr.14: 95433293-95433293 on Build GRCh38
rs770458132 Chr.14: 95433353-95433353 on Build GRCh38
rs779223333 Chr.14: 95433352-95433352 on Build GRCh38
rs777908933 Chr.14: 95433342-95433342 on Build GRCh38
rs780827065 Chr.14: 95433233-95433233 on Build GRCh38
rs373985574 Chr.14: 95433341-95433341 on Build GRCh38
rs201322570 Chr.14: 95433255-95433255 on Build GRCh38
rs763830611 Chr.14: 95433319-95433319 on Build GRCh38
rs761615747 Chr.14: 95433403-95433403 on Build GRCh38
rs889490423 Chr.14: 95433355-95433355 on Build GRCh38
rs756779761 Chr.14: 95433223-95433223 on Build GRCh38
rs200719423 Chr.14: 95433432-95433432 on Build GRCh38
rs777099004 Chr.14: 95433299-95433299 on Build GRCh38
rs199512224 Chr.14: 95433313-95433313 on Build GRCh38
rs746417592 Chr.14: 95433238-95433238 on Build GRCh38
rs780523611 Chr.14: 95433259-95433259 on Build GRCh38
rs35797947 Chr.14: 95433277-95433277 on Build GRCh38
rs751305624 Chr.14: 95433330-95433330 on Build GRCh38
rs747136131 Chr.14: 95433438-95433438 on Build GRCh38
rs201452286 Chr.14: 95433427-95433427 on Build GRCh38
rs756574233 Chr.14: 95433242-95433242 on Build GRCh38
rs767202552 Chr.14: 95433333-95433333 on Build GRCh38
rs768377239 Chr.14: 95433389-95433389 on Build GRCh38
rs776026362 Chr.14: 95433392-95433400 on Build GRCh38
rs746997603 Chr.14: 95433369-95433369 on Build GRCh38
rs749955924 Chr.14: 95433414-95433414 on Build GRCh38
rs182771300 Chr.14: 95433221-95433221 on Build GRCh38
rs759531353 Chr.14: 95433288-95433288 on Build GRCh38
rs116642867 Chr.14: 95433273-95433273 on Build GRCh38
rs939347179 Chr.14: 95433382-95433382 on Build GRCh38
rs140803384 Chr.14: 95433332-95433332 on Build GRCh38
rs372688254 Chr.14: 95433437-95433437 on Build GRCh38
rs12895537 Chr.14: 95433436-95433436 on Build GRCh38
rs1056558059 Chr.14: 95433385-95433385 on Build GRCh38
rs749555802 Chr.14: 95433266-95433266 on Build GRCh38
rs760060634 Chr.14: 95433420-95433420 on Build GRCh38
rs17092216 Chr.14: 95433357-95433357 on Build GRCh38
rs754346632 Chr.14: 95433426-95433426 on Build GRCh38
rs370756236 Chr.14: 95433349-95433349 on Build GRCh38
rs763588071 Chr.14: 95433218-95433218 on Build GRCh38
rs926461770 Chr.14: 95433269-95433269 on Build GRCh38
rs767219231 Chr.14: 95433409-95433409 on Build GRCh38
rs775243365 Chr.14: 95433378-95433378 on Build GRCh38
rs367936518 Chr.14: 95433380-95433380 on Build GRCh38
rs772184671 Chr.14: 95433283-95433283 on Build GRCh38
rs566836581 Chr.14: 95433350-95433350 on Build GRCh38

View Full Product Details