Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 64633823-64634094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACHD1
Entrez Gene ID
57685
Gene Name
cache domain containing 1
Gene Aliases
-
UniGene
Hs.443891
Gene Chromosome Location
Chr.1: 64470793-64693058 on Build GRCh38
Transcripts
NM_001293274.1 NM_020925.2 XM_011541862.1 XM_011541863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2238943 c.687G>A p.Q229Q Chr.1: 64634094-64634094 on Build GRCh38
6617286 c.797T>C p.V266A Chr.1: 64634051-64634051 on Build GRCh38
6617288 c.643G>C p.V215L Chr.1: 64634050-64634050 on Build GRCh38
5381368 c.669G>A p.R223R Chr.1: 64634076-64634076 on Build GRCh38
6617289 c.796G>C p.V266L Chr.1: 64634050-64634050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780957607 Chr.1: 64634019-64634019 on Build GRCh38
rs75667111 Chr.1: 64634033-64634033 on Build GRCh38
rs755551827 Chr.1: 64634039-64634039 on Build GRCh38
rs766546973 Chr.1: 64634021-64634021 on Build GRCh38
rs777372471 Chr.1: 64634017-64634018 on Build GRCh38
rs201033934 Chr.1: 64633996-64633996 on Build GRCh38
rs188597931 Chr.1: 64634073-64634073 on Build GRCh38
rs76989668 Chr.1: 64634017-64634017 on Build GRCh38
rs183370296 Chr.1: 64633868-64633868 on Build GRCh38
rs768259419 Chr.1: 64634070-64634070 on Build GRCh38
rs770293662 Chr.1: 64633914-64633914 on Build GRCh38
rs760997002 Chr.1: 64634016-64634017 on Build GRCh38
rs5774714 Chr.1: 64634018-64634018 on Build GRCh38
rs985480642 Chr.1: 64633984-64633984 on Build GRCh38
rs540823915 Chr.1: 64633879-64633879 on Build GRCh38
rs748771715 Chr.1: 64634069-64634069 on Build GRCh38
rs796286774 Chr.1: 64634018-64634019 on Build GRCh38
rs12044863 Chr.1: 64634065-64634065 on Build GRCh38
rs371026074 Chr.1: 64634074-64634074 on Build GRCh38
rs373496679 Chr.1: 64634017-64634017 on Build GRCh38
rs1045626980 Chr.1: 64633971-64633972 on Build GRCh38
rs762197555 Chr.1: 64634001-64634001 on Build GRCh38
rs200393627 Chr.1: 64633997-64633997 on Build GRCh38
rs1013217274 Chr.1: 64633905-64633905 on Build GRCh38
rs369697819 Chr.1: 64634033-64634033 on Build GRCh38
rs12044847 Chr.1: 64634043-64634043 on Build GRCh38
rs199662167 Chr.1: 64634007-64634007 on Build GRCh38
rs753972016 Chr.1: 64634025-64634025 on Build GRCh38
rs989298845 Chr.1: 64633902-64633902 on Build GRCh38
rs530434842 Chr.1: 64634071-64634071 on Build GRCh38
rs749016763 Chr.1: 64633980-64633980 on Build GRCh38
rs757134823 Chr.1: 64634037-64634037 on Build GRCh38
rs751276734 Chr.1: 64634018-64634020 on Build GRCh38
rs918999288 Chr.1: 64633867-64633867 on Build GRCh38
rs781264766 Chr.1: 64634038-64634038 on Build GRCh38
rs559149818 Chr.1: 64633983-64633983 on Build GRCh38
rs757583949 Chr.1: 64633929-64633930 on Build GRCh38
rs1030854090 Chr.1: 64633855-64633855 on Build GRCh38
rs912517743 Chr.1: 64633986-64633986 on Build GRCh38
rs764180229 Chr.1: 64634018-64634019 on Build GRCh38
rs894322481 Chr.1: 64633890-64633890 on Build GRCh38
rs760657377 Chr.1: 64634019-64634019 on Build GRCh38
rs931882929 Chr.1: 64633970-64633970 on Build GRCh38
rs751356579 Chr.1: 64634036-64634036 on Build GRCh38
rs775346734 Chr.1: 64633857-64633857 on Build GRCh38
rs80072687 Chr.1: 64634032-64634032 on Build GRCh38
rs866835503 Chr.1: 64634018-64634018 on Build GRCh38
rs921878728 Chr.1: 64633963-64633963 on Build GRCh38
rs773781156 Chr.1: 64634014-64634014 on Build GRCh38
rs762253626 Chr.1: 64633888-64633888 on Build GRCh38

View Full Product Details