Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177508280-177508513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOK3
Entrez Gene ID
79930
Gene Name
docking protein 3
Gene Aliases
DOKL
UniGene
Hs.720849
Gene Chromosome Location
Chr.5: 177501904-177510507 on Build GRCh38
Transcripts
NM_001144875.1 NM_001144876.1 NM_001308235.1 NM_001308236.1 NM_024872.2 XM_005265985.3 XM_005265987.1 XM_011534654.2 XM_011534655.2 XM_011534656.2 XM_011534657.2 XM_017009883.1 XM_017009884.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1436351 c.107G>A p.G36D Chr.5: 177508502-177508502 on Build GRCh38
6378566 c.371A>T p.E124V Chr.5: 177508406-177508406 on Build GRCh38
1436350 c.275G>A p.G92D Chr.5: 177508502-177508502 on Build GRCh38
6378128 c.461C>A p.T154N Chr.5: 177508316-177508316 on Build GRCh38
1436348 c.314G>A p.R105Q Chr.5: 177508463-177508463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752829173 Chr.5: 177508356-177508356 on Build GRCh38
rs375564754 Chr.5: 177508308-177508308 on Build GRCh38
rs778804038 Chr.5: 177508325-177508325 on Build GRCh38
rs760143673 Chr.5: 177508349-177508349 on Build GRCh38
rs778697790 Chr.5: 177508363-177508363 on Build GRCh38
rs754060367 Chr.5: 177508360-177508360 on Build GRCh38
rs768097645 Chr.5: 177508392-177508392 on Build GRCh38
rs745849199 Chr.5: 177508329-177508329 on Build GRCh38
rs565362873 Chr.5: 177508345-177508345 on Build GRCh38
rs780588537 Chr.5: 177508474-177508474 on Build GRCh38
rs535291002 Chr.5: 177508429-177508429 on Build GRCh38
rs754470446 Chr.5: 177508465-177508465 on Build GRCh38
rs746514207 Chr.5: 177508384-177508384 on Build GRCh38
rs111533164 Chr.5: 177508332-177508332 on Build GRCh38
rs964755829 Chr.5: 177508327-177508327 on Build GRCh38
rs144620098 Chr.5: 177508372-177508372 on Build GRCh38
rs774869740 Chr.5: 177508412-177508412 on Build GRCh38
rs544789441 Chr.5: 177508492-177508492 on Build GRCh38
rs776316205 Chr.5: 177508393-177508393 on Build GRCh38
rs372411602 Chr.5: 177508415-177508415 on Build GRCh38
rs949192537 Chr.5: 177508354-177508354 on Build GRCh38
rs148416579 Chr.5: 177508333-177508333 on Build GRCh38
rs900493543 Chr.5: 177508460-177508460 on Build GRCh38
rs767648183 Chr.5: 177508353-177508353 on Build GRCh38
rs374999538 Chr.5: 177508450-177508450 on Build GRCh38
rs146755459 Chr.5: 177508416-177508416 on Build GRCh38
rs777637154 Chr.5: 177508485-177508485 on Build GRCh38
rs747745711 Chr.5: 177508477-177508477 on Build GRCh38
rs926937930 Chr.5: 177508369-177508369 on Build GRCh38
rs369855241 Chr.5: 177508335-177508335 on Build GRCh38
rs369375986 Chr.5: 177508461-177508461 on Build GRCh38
rs758334169 Chr.5: 177508371-177508371 on Build GRCh38
rs887828416 Chr.5: 177508309-177508309 on Build GRCh38
rs762031427 Chr.5: 177508430-177508430 on Build GRCh38
rs912547032 Chr.5: 177508397-177508397 on Build GRCh38
rs940016093 Chr.5: 177508385-177508385 on Build GRCh38
rs753928803 Chr.5: 177508334-177508335 on Build GRCh38
rs372201708 Chr.5: 177508305-177508305 on Build GRCh38
rs771374795 Chr.5: 177508403-177508403 on Build GRCh38
rs771518601 Chr.5: 177508331-177508331 on Build GRCh38
rs750211007 Chr.5: 177508442-177508442 on Build GRCh38
rs747833593 Chr.5: 177508399-177508399 on Build GRCh38
rs116453223 Chr.5: 177508441-177508441 on Build GRCh38
rs150922665 Chr.5: 177508341-177508341 on Build GRCh38
rs758932270 Chr.5: 177508463-177508463 on Build GRCh38
rs1021192837 Chr.5: 177508464-177508464 on Build GRCh38
rs764124229 Chr.5: 177508423-177508423 on Build GRCh38
rs1018000249 Chr.5: 177508324-177508324 on Build GRCh38
rs756373791 Chr.5: 177508301-177508301 on Build GRCh38
rs147721747 Chr.5: 177508312-177508312 on Build GRCh38
rs769576036 Chr.5: 177508480-177508480 on Build GRCh38
rs540258218 Chr.5: 177508339-177508339 on Build GRCh38
rs774957822 Chr.5: 177508346-177508346 on Build GRCh38
rs142573859 Chr.5: 177508340-177508340 on Build GRCh38
rs760055646 Chr.5: 177508413-177508413 on Build GRCh38
rs372954918 Chr.5: 177508486-177508486 on Build GRCh38
rs755247599 Chr.5: 177508374-177508375 on Build GRCh38
rs1048324258 Chr.5: 177508424-177508424 on Build GRCh38
rs145411390 Chr.5: 177508484-177508484 on Build GRCh38
rs756218542 Chr.5: 177508357-177508357 on Build GRCh38
rs757018092 Chr.5: 177508362-177508362 on Build GRCh38

View Full Product Details