Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 30696289-30696562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSR
Entrez Gene ID
2936
Gene Name
glutathione-disulfide reductase
Gene Aliases
HEL-75, HEL-S-122m
UniGene
Hs.271510
Gene Chromosome Location
Chr.8: 30678061-30727969 on Build GRCh38
Transcripts
NM_000637.3 NM_001195102.1 NM_001195103.1 NM_001195104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6316707 c.702C>A p.S234R Chr.8: 30696473-30696473 on Build GRCh38
3982507 c.696-10C>T p.? Chr.8: 30696489-30696489 on Build GRCh38
5874560 c.763T>C p.S255P Chr.8: 30696412-30696412 on Build GRCh38
254719 c.752C>G p.S251* Chr.8: 30696423-30696423 on Build GRCh38
1099120 c.703G>A p.V235I Chr.8: 30696472-30696472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376634766 Chr.8: 30696486-30696486 on Build GRCh38
rs745674409 Chr.8: 30696390-30696390 on Build GRCh38
rs140665617 Chr.8: 30696391-30696391 on Build GRCh38
rs755033052 Chr.8: 30696332-30696332 on Build GRCh38
rs8190996 Chr.8: 30696489-30696489 on Build GRCh38
rs368906548 Chr.8: 30696403-30696403 on Build GRCh38
rs756475234 Chr.8: 30696472-30696472 on Build GRCh38
rs145851500 Chr.8: 30696478-30696478 on Build GRCh38
rs756689722 Chr.8: 30696398-30696398 on Build GRCh38
rs374500135 Chr.8: 30696427-30696427 on Build GRCh38
rs773603845 Chr.8: 30696416-30696416 on Build GRCh38
rs779889177 Chr.8: 30696482-30696482 on Build GRCh38
rs778911940 Chr.8: 30696387-30696387 on Build GRCh38
rs750234827 Chr.8: 30696477-30696477 on Build GRCh38
rs767650258 Chr.8: 30696469-30696469 on Build GRCh38
rs2738991 Chr.8: 30696507-30696507 on Build GRCh38
rs757135199 Chr.8: 30696386-30696386 on Build GRCh38
rs200738734 Chr.8: 30696401-30696401 on Build GRCh38
rs768087560 Chr.8: 30696404-30696404 on Build GRCh38
rs767879816 Chr.8: 30696340-30696340 on Build GRCh38
rs751636803 Chr.8: 30696330-30696330 on Build GRCh38
rs771257873 Chr.8: 30696502-30696502 on Build GRCh38
rs1055981830 Chr.8: 30696359-30696359 on Build GRCh38
rs769482085 Chr.8: 30696496-30696496 on Build GRCh38
rs759541499 Chr.8: 30696456-30696456 on Build GRCh38
rs8190997 Chr.8: 30696394-30696394 on Build GRCh38
rs774878841 Chr.8: 30696451-30696451 on Build GRCh38
rs568529653 Chr.8: 30696488-30696488 on Build GRCh38
rs200120959 Chr.8: 30696420-30696420 on Build GRCh38
rs754893920 Chr.8: 30696487-30696487 on Build GRCh38
rs867397746 Chr.8: 30696428-30696428 on Build GRCh38
rs369812470 Chr.8: 30696356-30696356 on Build GRCh38
rs769431245 Chr.8: 30696414-30696414 on Build GRCh38
rs1027270872 Chr.8: 30696453-30696453 on Build GRCh38
rs140455984 Chr.8: 30696473-30696473 on Build GRCh38
rs753883260 Chr.8: 30696423-30696423 on Build GRCh38
rs760149430 Chr.8: 30696521-30696521 on Build GRCh38
rs376658099 Chr.8: 30696405-30696405 on Build GRCh38
rs779437569 Chr.8: 30696513-30696513 on Build GRCh38
rs772868107 Chr.8: 30696497-30696497 on Build GRCh38
rs961655015 Chr.8: 30696476-30696476 on Build GRCh38
rs749644111 Chr.8: 30696501-30696501 on Build GRCh38

View Full Product Details