Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 126306521-126306792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCPS
Entrez Gene ID
28960
Gene Name
decapping enzyme, scavenger
Gene Aliases
ARS, DCS1, HINT-5, HINT5, HSL1, HSPC015
UniGene
Hs.504249
Gene Chromosome Location
Chr.11: 126303301-126345754 on Build GRCh38
Transcripts
NM_014026.4 XM_011542778.2 XM_017017612.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
355840 c.261G>T p.T87T Chr.11: 126306629-126306629 on Build GRCh38
7118441 c.294G>A p.L98L Chr.11: 126306662-126306662 on Build GRCh38
4409631 c.318G>A p.L106L Chr.11: 126306686-126306686 on Build GRCh38
415275 c.241G>A p.V81I Chr.11: 126306609-126306609 on Build GRCh38
6204005 c.274G>A p.E92K Chr.11: 126306642-126306642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765762962 Chr.11: 126306765-126306766 on Build GRCh38
rs141856350 Chr.11: 126306664-126306664 on Build GRCh38
rs145173937 Chr.11: 126306589-126306589 on Build GRCh38
rs148042649 Chr.11: 126306698-126306698 on Build GRCh38
rs939531956 Chr.11: 126306725-126306725 on Build GRCh38
rs992770994 Chr.11: 126306755-126306755 on Build GRCh38
rs769591791 Chr.11: 126306580-126306580 on Build GRCh38
rs768792009 Chr.11: 126306719-126306719 on Build GRCh38
rs372937832 Chr.11: 126306563-126306563 on Build GRCh38
rs764625589 Chr.11: 126306549-126306551 on Build GRCh38
rs139850206 Chr.11: 126306632-126306632 on Build GRCh38
rs751972143 Chr.11: 126306641-126306641 on Build GRCh38
rs149150998 Chr.11: 126306595-126306595 on Build GRCh38
rs759338894 Chr.11: 126306712-126306712 on Build GRCh38
rs935312176 Chr.11: 126306726-126306726 on Build GRCh38
rs762192163 Chr.11: 126306731-126306731 on Build GRCh38
rs760773746 Chr.11: 126306607-126306607 on Build GRCh38
rs750867339 Chr.11: 126306752-126306752 on Build GRCh38
rs1008367682 Chr.11: 126306603-126306603 on Build GRCh38
rs11557735 Chr.11: 126306586-126306586 on Build GRCh38
rs201095573 Chr.11: 126306583-126306584 on Build GRCh38
rs138737928 Chr.11: 126306628-126306628 on Build GRCh38
rs762947260 Chr.11: 126306593-126306593 on Build GRCh38
rs182840030 Chr.11: 126306672-126306672 on Build GRCh38
rs774374339 Chr.11: 126306707-126306707 on Build GRCh38
rs762065785 Chr.11: 126306616-126306616 on Build GRCh38
rs776782460 Chr.11: 126306720-126306720 on Build GRCh38
rs868861146 Chr.11: 126306682-126306682 on Build GRCh38
rs371538904 Chr.11: 126306596-126306596 on Build GRCh38
rs750901619 Chr.11: 126306621-126306621 on Build GRCh38
rs935428244 Chr.11: 126306582-126306582 on Build GRCh38
rs1014656265 Chr.11: 126306656-126306656 on Build GRCh38
rs533266199 Chr.11: 126306629-126306629 on Build GRCh38
rs377290056 Chr.11: 126306567-126306567 on Build GRCh38
rs375853628 Chr.11: 126306697-126306697 on Build GRCh38
rs188890331 Chr.11: 126306684-126306684 on Build GRCh38
rs202227377 Chr.11: 126306754-126306754 on Build GRCh38
rs960834180 Chr.11: 126306659-126306659 on Build GRCh38
rs637462 Chr.11: 126306683-126306683 on Build GRCh38
rs754539521 Chr.11: 126306650-126306650 on Build GRCh38
rs749063895 Chr.11: 126306687-126306687 on Build GRCh38
rs144876585 Chr.11: 126306608-126306608 on Build GRCh38
rs765540619 Chr.11: 126306735-126306735 on Build GRCh38
rs754132367 Chr.11: 126306609-126306609 on Build GRCh38

View Full Product Details