Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49253009-49253262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXP3
Entrez Gene ID
50943
Gene Name
forkhead box P3
Gene Aliases
AIID, DIETER, IPEX, JM2, PIDX, XPID
UniGene
Hs.247700
Gene Chromosome Location
Chr.X: 49250436-49266505 on Build GRCh38
Transcripts
NM_001114377.1 NM_014009.3 XM_006724533.2 XM_011543916.2 XM_017029565.1 XM_017029566.1 XM_017029567.1

SNP Detail

SNP ID Chromosome Location
rs192502436 Chr.X: 49253058-49253058 on Build GRCh38
rs782472240 Chr.X: 49253153-49253153 on Build GRCh38
rs782523136 Chr.X: 49253225-49253225 on Build GRCh38
rs782368278 Chr.X: 49253219-49253219 on Build GRCh38
rs782694089 Chr.X: 49253110-49253110 on Build GRCh38
rs781905807 Chr.X: 49253138-49253138 on Build GRCh38
rs782539207 Chr.X: 49253139-49253139 on Build GRCh38
rs11465478 Chr.X: 49253109-49253109 on Build GRCh38
rs782368010 Chr.X: 49253215-49253215 on Build GRCh38
rs122467173 Chr.X: 49253200-49253200 on Build GRCh38
rs1057517736 Chr.X: 49253161-49253161 on Build GRCh38
rs374651495 Chr.X: 49253144-49253144 on Build GRCh38
rs782475933 Chr.X: 49253214-49253214 on Build GRCh38
rs782429831 Chr.X: 49253193-49253193 on Build GRCh38
rs72650694 Chr.X: 49253095-49253095 on Build GRCh38
rs1009561176 Chr.X: 49253041-49253041 on Build GRCh38
rs782092269 Chr.X: 49253234-49253234 on Build GRCh38
rs781826347 Chr.X: 49253103-49253103 on Build GRCh38
rs79563992 Chr.X: 49253213-49253213 on Build GRCh38
rs371301543 Chr.X: 49253195-49253195 on Build GRCh38
rs367860281 Chr.X: 49253186-49253186 on Build GRCh38
rs782649847 Chr.X: 49253143-49253143 on Build GRCh38
rs77362352 Chr.X: 49253078-49253078 on Build GRCh38
rs782255694 Chr.X: 49253209-49253209 on Build GRCh38
rs886044787 Chr.X: 49253155-49253155 on Build GRCh38
rs782591493 Chr.X: 49253175-49253175 on Build GRCh38

View Full Product Details