Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38466173-38466422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP23
Entrez Gene ID
57636
Gene Name
Rho GTPase activating protein 23
Gene Aliases
-
UniGene
Hs.374446
Gene Chromosome Location
Chr.17: 38419286-38512392 on Build GRCh38
Transcripts
NM_001199417.1 XM_006721991.1 XM_006721992.3 XM_006721993.3 XM_006721994.3 XM_011525073.1 XM_011525074.2 XM_011525075.2 XM_011525076.2 XM_011525077.2 XM_017024902.1

SNP Detail

SNP ID Chromosome Location
rs889384455 Chr.17: 38466225-38466225 on Build GRCh38
rs964200325 Chr.17: 38466297-38466297 on Build GRCh38
rs778025028 Chr.17: 38466271-38466271 on Build GRCh38
rs751834913 Chr.17: 38466221-38466221 on Build GRCh38
rs542426232 Chr.17: 38466240-38466240 on Build GRCh38
rs368977450 Chr.17: 38466282-38466282 on Build GRCh38
rs759925670 Chr.17: 38466396-38466396 on Build GRCh38
rs758815578 Chr.17: 38466204-38466204 on Build GRCh38
rs528267224 Chr.17: 38466281-38466281 on Build GRCh38
rs758609941 Chr.17: 38466243-38466243 on Build GRCh38
rs949656082 Chr.17: 38466374-38466374 on Build GRCh38
rs868459145 Chr.17: 38466296-38466296 on Build GRCh38
rs768764279 Chr.17: 38466325-38466325 on Build GRCh38
rs774256741 Chr.17: 38466341-38466341 on Build GRCh38
rs8075324 Chr.17: 38466241-38466241 on Build GRCh38
rs879449507 Chr.17: 38466350-38466350 on Build GRCh38
rs1023284103 Chr.17: 38466343-38466343 on Build GRCh38
rs573835627 Chr.17: 38466222-38466222 on Build GRCh38
rs755544986 Chr.17: 38466235-38466235 on Build GRCh38
rs577820928 Chr.17: 38466393-38466393 on Build GRCh38
rs765771265 Chr.17: 38466392-38466392 on Build GRCh38
rs760162545 Chr.17: 38466199-38466199 on Build GRCh38
rs974554362 Chr.17: 38466319-38466319 on Build GRCh38
rs369360815 Chr.17: 38466367-38466367 on Build GRCh38
rs760382401 Chr.17: 38466375-38466375 on Build GRCh38
rs192671655 Chr.17: 38466366-38466366 on Build GRCh38
rs371590163 Chr.17: 38466347-38466347 on Build GRCh38
rs1027163396 Chr.17: 38466339-38466339 on Build GRCh38
rs898026369 Chr.17: 38466269-38466269 on Build GRCh38
rs925588988 Chr.17: 38466193-38466193 on Build GRCh38
rs994287605 Chr.17: 38466277-38466277 on Build GRCh38
rs956575828 Chr.17: 38466370-38466370 on Build GRCh38
rs1045904929 Chr.17: 38466194-38466194 on Build GRCh38
rs750576576 Chr.17: 38466268-38466268 on Build GRCh38
rs960627054 Chr.17: 38466337-38466337 on Build GRCh38
rs749130674 Chr.17: 38466303-38466303 on Build GRCh38
rs375565906 Chr.17: 38466272-38466272 on Build GRCh38
rs1010119823 Chr.17: 38466264-38466264 on Build GRCh38
rs62074684 Chr.17: 38466205-38466205 on Build GRCh38
rs1008255072 Chr.17: 38466273-38466273 on Build GRCh38

View Full Product Details