Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32813718-32813943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DOB
Entrez Gene ID
3112
Gene Name
major histocompatibility complex, class II, DO beta
Gene Aliases
DOB
UniGene
Hs.1802
Gene Chromosome Location
Chr.6: 32812763-32817048 on Build GRCh38
Transcripts
NM_002120.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6431110 c.707T>C p.L236P Chr.6: 32813770-32813770 on Build GRCh38
5666716 c.644G>T p.R215I Chr.6: 32813833-32813833 on Build GRCh38
3625493 c.725G>A p.G242E Chr.6: 32813752-32813752 on Build GRCh38
3625494 c.703G>A p.G235R Chr.6: 32813774-32813774 on Build GRCh38
5414325 c.697C>T p.L233L Chr.6: 32813780-32813780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112802855 Chr.6: 32813776-32813776 on Build GRCh38
rs373364805 Chr.6: 32813799-32813799 on Build GRCh38
rs369377287 Chr.6: 32813788-32813788 on Build GRCh38
rs2070121 Chr.6: 32813777-32813777 on Build GRCh38
rs2071478 Chr.6: 32813755-32813755 on Build GRCh38
rs376049376 Chr.6: 32813809-32813809 on Build GRCh38
rs558387026 Chr.6: 32813748-32813748 on Build GRCh38
rs985256873 Chr.6: 32813850-32813850 on Build GRCh38
rs769104615 Chr.6: 32813769-32813769 on Build GRCh38
rs974898938 Chr.6: 32813791-32813791 on Build GRCh38
rs770667884 Chr.6: 32813811-32813811 on Build GRCh38
rs752575374 Chr.6: 32813883-32813883 on Build GRCh38
rs994997952 Chr.6: 32813908-32813908 on Build GRCh38
rs1029193392 Chr.6: 32813830-32813830 on Build GRCh38
rs1036440494 Chr.6: 32813800-32813800 on Build GRCh38
rs778741202 Chr.6: 32813832-32813832 on Build GRCh38
rs759234434 Chr.6: 32813847-32813847 on Build GRCh38
rs111971502 Chr.6: 32813746-32813746 on Build GRCh38
rs966321746 Chr.6: 32813778-32813778 on Build GRCh38
rs865838330 Chr.6: 32813812-32813812 on Build GRCh38
rs745752752 Chr.6: 32813844-32813844 on Build GRCh38
rs780341623 Chr.6: 32813756-32813756 on Build GRCh38
rs765140881 Chr.6: 32813855-32813855 on Build GRCh38
rs940653619 Chr.6: 32813874-32813874 on Build GRCh38
rs772792120 Chr.6: 32813802-32813802 on Build GRCh38
rs2621330 Chr.6: 32813747-32813747 on Build GRCh38
rs55816512 Chr.6: 32813763-32813763 on Build GRCh38
rs201921473 Chr.6: 32813779-32813779 on Build GRCh38

View Full Product Details