Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27463011-27463258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA2
Entrez Gene ID
1135
Gene Name
cholinergic receptor nicotinic alpha 2 subunit
Gene Aliases
-
UniGene
Hs.57718
Gene Chromosome Location
Chr.8: 27459761-27479296 on Build GRCh38
Transcripts
NM_000742.3 NM_001282455.1 XM_005273397.2 XM_006716282.1 XM_011544388.1 XM_011544389.1 XM_017013003.1 XM_017013004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3899287 c.1278A>G p.A426A Chr.8: 27463165-27463165 on Build GRCh38
3899286 c.1358C>T p.A453V Chr.8: 27463085-27463085 on Build GRCh38
4981278 c.1282C>T p.H428Y Chr.8: 27463161-27463161 on Build GRCh38
5503353 c.1194C>T p.S398S Chr.8: 27463249-27463249 on Build GRCh38
1201120 c.1217G>A p.S406N Chr.8: 27463226-27463226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770893077 Chr.8: 27463229-27463229 on Build GRCh38
rs963988264 Chr.8: 27463031-27463031 on Build GRCh38
rs747909980 Chr.8: 27463220-27463220 on Build GRCh38
rs528317928 Chr.8: 27463072-27463072 on Build GRCh38
rs764626569 Chr.8: 27463191-27463191 on Build GRCh38
rs796712573 Chr.8: 27463082-27463082 on Build GRCh38
rs751242542 Chr.8: 27463070-27463070 on Build GRCh38
rs367771198 Chr.8: 27463222-27463222 on Build GRCh38
rs778197366 Chr.8: 27463089-27463089 on Build GRCh38
rs976672656 Chr.8: 27463069-27463069 on Build GRCh38
rs866577498 Chr.8: 27463140-27463140 on Build GRCh38
rs760956524 Chr.8: 27463189-27463189 on Build GRCh38
rs56298562 Chr.8: 27463119-27463119 on Build GRCh38
rs377612702 Chr.8: 27463058-27463058 on Build GRCh38
rs530383631 Chr.8: 27463168-27463168 on Build GRCh38
rs753041128 Chr.8: 27463074-27463074 on Build GRCh38
rs868846472 Chr.8: 27463199-27463199 on Build GRCh38
rs756635424 Chr.8: 27463078-27463078 on Build GRCh38
rs377524925 Chr.8: 27463111-27463111 on Build GRCh38
rs1021964888 Chr.8: 27463169-27463169 on Build GRCh38
rs370957296 Chr.8: 27463075-27463075 on Build GRCh38
rs746067361 Chr.8: 27463109-27463109 on Build GRCh38
rs753892989 Chr.8: 27463091-27463091 on Build GRCh38
rs751842443 Chr.8: 27463150-27463150 on Build GRCh38
rs939566379 Chr.8: 27463228-27463228 on Build GRCh38
rs770447673 Chr.8: 27463151-27463151 on Build GRCh38
rs772465304 Chr.8: 27463112-27463112 on Build GRCh38
rs968676056 Chr.8: 27463203-27463203 on Build GRCh38
rs767138285 Chr.8: 27463173-27463173 on Build GRCh38
rs868127322 Chr.8: 27463200-27463200 on Build GRCh38
rs762463166 Chr.8: 27463062-27463062 on Build GRCh38
rs757019076 Chr.8: 27463071-27463071 on Build GRCh38
rs772765800 Chr.8: 27463050-27463050 on Build GRCh38
rs777716357 Chr.8: 27463223-27463223 on Build GRCh38
rs138721954 Chr.8: 27463166-27463166 on Build GRCh38
rs750599711 Chr.8: 27463207-27463207 on Build GRCh38
rs373599393 Chr.8: 27463198-27463198 on Build GRCh38
rs370878600 Chr.8: 27463195-27463195 on Build GRCh38
rs765154757 Chr.8: 27463042-27463042 on Build GRCh38
rs762408022 Chr.8: 27463142-27463142 on Build GRCh38
rs78561863 Chr.8: 27463204-27463204 on Build GRCh38
rs765948265 Chr.8: 27463065-27463065 on Build GRCh38
rs753088694 Chr.8: 27463178-27463178 on Build GRCh38
rs747952090 Chr.8: 27463124-27463124 on Build GRCh38
rs754422429 Chr.8: 27463193-27463193 on Build GRCh38
rs764302114 Chr.8: 27463116-27463116 on Build GRCh38
rs544902151 Chr.8: 27463108-27463108 on Build GRCh38
rs775578495 Chr.8: 27463126-27463126 on Build GRCh38
rs771994243 Chr.8: 27463221-27463221 on Build GRCh38
rs141721605 Chr.8: 27463209-27463209 on Build GRCh38
rs773026258 Chr.8: 27463141-27463141 on Build GRCh38
rs749199596 Chr.8: 27463225-27463225 on Build GRCh38
rs374281427 Chr.8: 27463100-27463100 on Build GRCh38

View Full Product Details