Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62573704-62573971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF1G
Entrez Gene ID
1937
Gene Name
eukaryotic translation elongation factor 1 gamma
Gene Aliases
EF1G, GIG35
UniGene
Hs.144835
Gene Chromosome Location
Chr.11: 62559601-62573988 on Build GRCh38
Transcripts
NM_001404.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7211750 c.9T>C p.A3A Chr.11: 62573834-62573834 on Build GRCh38
116325 c.10G>T p.G4W Chr.11: 62573833-62573833 on Build GRCh38
1194221 c.11G>T p.G4V Chr.11: 62573832-62573832 on Build GRCh38
2038855 c.6G>T p.A2A Chr.11: 62573837-62573837 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs946120827 Chr.11: 62573816-62573816 on Build GRCh38
rs370202672 Chr.11: 62573863-62573863 on Build GRCh38
rs990816606 Chr.11: 62573918-62573918 on Build GRCh38
rs959234798 Chr.11: 62573751-62573751 on Build GRCh38
rs546962387 Chr.11: 62573819-62573819 on Build GRCh38
rs549104690 Chr.11: 62573914-62573914 on Build GRCh38
rs568878044 Chr.11: 62573904-62573904 on Build GRCh38
rs760577306 Chr.11: 62573861-62573861 on Build GRCh38
rs754034871 Chr.11: 62573787-62573787 on Build GRCh38
rs911456196 Chr.11: 62573740-62573740 on Build GRCh38
rs778306394 Chr.11: 62573815-62573815 on Build GRCh38
rs758152342 Chr.11: 62573798-62573800 on Build GRCh38
rs371074771 Chr.11: 62573795-62573795 on Build GRCh38
rs752095509 Chr.11: 62573866-62573866 on Build GRCh38
rs768353853 Chr.11: 62573838-62573838 on Build GRCh38
rs754553441 Chr.11: 62573885-62573885 on Build GRCh38
rs374485717 Chr.11: 62573817-62573817 on Build GRCh38
rs778668475 Chr.11: 62573886-62573886 on Build GRCh38
rs773116930 Chr.11: 62573924-62573924 on Build GRCh38
rs199701843 Chr.11: 62573797-62573797 on Build GRCh38
rs747927647 Chr.11: 62573843-62573843 on Build GRCh38
rs766407278 Chr.11: 62573783-62573783 on Build GRCh38
rs748941956 Chr.11: 62573829-62573829 on Build GRCh38
rs879717418 Chr.11: 62573758-62573758 on Build GRCh38
rs922161548 Chr.11: 62573878-62573878 on Build GRCh38
rs201889192 Chr.11: 62573876-62573876 on Build GRCh38
rs1008385350 Chr.11: 62573901-62573901 on Build GRCh38
rs928763167 Chr.11: 62573825-62573825 on Build GRCh38
rs964291383 Chr.11: 62573905-62573905 on Build GRCh38
rs771935705 Chr.11: 62573858-62573858 on Build GRCh38
rs758715719 Chr.11: 62573809-62573809 on Build GRCh38
rs777516967 Chr.11: 62573824-62573825 on Build GRCh38
rs758762019 Chr.11: 62573864-62573864 on Build GRCh38
rs977474284 Chr.11: 62573928-62573928 on Build GRCh38
rs201389774 Chr.11: 62573865-62573865 on Build GRCh38
rs914864286 Chr.11: 62573921-62573921 on Build GRCh38
rs955130326 Chr.11: 62573898-62573898 on Build GRCh38
rs748118824 Chr.11: 62573857-62573857 on Build GRCh38
rs376137209 Chr.11: 62573759-62573759 on Build GRCh38
rs760767592 Chr.11: 62573782-62573782 on Build GRCh38
rs3741241 Chr.11: 62573862-62573862 on Build GRCh38
rs1025394693 Chr.11: 62573894-62573894 on Build GRCh38
rs755262453 Chr.11: 62573789-62573789 on Build GRCh38
rs771793192 Chr.11: 62573852-62573852 on Build GRCh38
rs753001715 Chr.11: 62573803-62573803 on Build GRCh38
rs991933416 Chr.11: 62573793-62573793 on Build GRCh38
rs765450614 Chr.11: 62573800-62573800 on Build GRCh38
rs914562145 Chr.11: 62573798-62573798 on Build GRCh38
rs548928221 Chr.11: 62573897-62573897 on Build GRCh38
rs898163045 Chr.11: 62573879-62573879 on Build GRCh38
rs757768987 Chr.11: 62573867-62573867 on Build GRCh38
rs1033547224 Chr.11: 62573750-62573750 on Build GRCh38
rs781764888 Chr.11: 62573869-62573869 on Build GRCh38

View Full Product Details