Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41419078-41419345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCKDHA
Entrez Gene ID
593
Gene Name
branched chain keto acid dehydrogenase E1, alpha polypeptide
Gene Aliases
BCKDE1A, MSU, MSUD1, OVD1A
UniGene
Hs.433307
Gene Chromosome Location
Chr.19: 41397789-41425005 on Build GRCh38
Transcripts
NM_000709.3 NM_001164783.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3823182 c.506A>G p.Y169C Chr.19: 41419156-41419156 on Build GRCh38
7124597 c.492G>A p.L164L Chr.19: 41419142-41419142 on Build GRCh38
3227085 c.605T>A p.F202Y Chr.19: 41419255-41419255 on Build GRCh38
3227081 c.486T>C p.G162G Chr.19: 41419136-41419136 on Build GRCh38
997027 c.632C>T p.T211M Chr.19: 41419282-41419282 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001407346 Chr.19: 41419144-41419144 on Build GRCh38
rs745668397 Chr.19: 41419240-41419240 on Build GRCh38
rs767906298 Chr.19: 41419121-41419121 on Build GRCh38
rs377517768 Chr.19: 41419150-41419150 on Build GRCh38
rs754613642 Chr.19: 41419157-41419157 on Build GRCh38
rs572679160 Chr.19: 41419099-41419099 on Build GRCh38
rs750857655 Chr.19: 41419154-41419154 on Build GRCh38
rs370068964 Chr.19: 41419256-41419256 on Build GRCh38
rs759832847 Chr.19: 41419115-41419115 on Build GRCh38
rs780047475 Chr.19: 41419322-41419322 on Build GRCh38
rs973936708 Chr.19: 41419295-41419295 on Build GRCh38
rs770609569 Chr.19: 41419106-41419106 on Build GRCh38
rs34956071 Chr.19: 41419158-41419158 on Build GRCh38
rs772349835 Chr.19: 41419247-41419247 on Build GRCh38
rs775651033 Chr.19: 41419122-41419122 on Build GRCh38
rs762226416 Chr.19: 41419275-41419275 on Build GRCh38
rs144995574 Chr.19: 41419268-41419268 on Build GRCh38
rs764618560 Chr.19: 41419142-41419142 on Build GRCh38
rs772012508 Chr.19: 41419318-41419318 on Build GRCh38
rs988176395 Chr.19: 41419257-41419257 on Build GRCh38
rs755611354 Chr.19: 41419178-41419178 on Build GRCh38
rs963488715 Chr.19: 41419294-41419295 on Build GRCh38
rs752322921 Chr.19: 41419298-41419298 on Build GRCh38
rs767118332 Chr.19: 41419291-41419291 on Build GRCh38
rs768600012 Chr.19: 41419265-41419265 on Build GRCh38
rs750729027 Chr.19: 41419283-41419283 on Build GRCh38
rs767888099 Chr.19: 41419321-41419321 on Build GRCh38
rs904274723 Chr.19: 41419117-41419117 on Build GRCh38
rs369477231 Chr.19: 41419317-41419317 on Build GRCh38
rs114178789 Chr.19: 41419107-41419107 on Build GRCh38
rs201333801 Chr.19: 41419309-41419309 on Build GRCh38
rs10404506 Chr.19: 41419289-41419289 on Build GRCh38
rs981398635 Chr.19: 41419199-41419199 on Build GRCh38
rs398123502 Chr.19: 41419149-41419149 on Build GRCh38
rs781632532 Chr.19: 41419131-41419131 on Build GRCh38
rs201537906 Chr.19: 41419218-41419218 on Build GRCh38
rs754327984 Chr.19: 41419145-41419145 on Build GRCh38
rs925562568 Chr.19: 41419207-41419207 on Build GRCh38
rs575806482 Chr.19: 41419235-41419235 on Build GRCh38
rs886042356 Chr.19: 41419215-41419215 on Build GRCh38
rs747591833 Chr.19: 41419167-41419167 on Build GRCh38
rs190610188 Chr.19: 41419248-41419248 on Build GRCh38
rs969597284 Chr.19: 41419153-41419153 on Build GRCh38
rs777391858 Chr.19: 41419217-41419217 on Build GRCh38
rs933412833 Chr.19: 41419300-41419300 on Build GRCh38
rs770967739 Chr.19: 41419226-41419226 on Build GRCh38
rs778995282 Chr.19: 41419312-41419312 on Build GRCh38
rs755828892 Chr.19: 41419303-41419303 on Build GRCh38
rs1002166107 Chr.19: 41419278-41419278 on Build GRCh38
rs398123503 Chr.19: 41419282-41419282 on Build GRCh38
rs760838451 Chr.19: 41419125-41419125 on Build GRCh38
rs762084007 Chr.19: 41419157-41419157 on Build GRCh38
rs777143993 Chr.19: 41419308-41419308 on Build GRCh38

View Full Product Details